Skip to Content
Merck
All Photos(2)

Key Documents

R2500

Sigma-Aldrich

all trans-Retinal

powder, ≥98%

Synonym(s):

Vitamin A aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

color

yellow

mp

62-64 °C

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(=O)\C=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,15H,7,10,14H2,1-5H3/b9-6+,12-11+,16-8+,17-13+

InChI key

NCYCYZXNIZJOKI-OVSJKPMPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

All trans-Retinal is one of the major derivatives of vitamin A group. A variety of food serves as a source of vitamin A. It is predominant in liver and among the brightly colored vegetables.

Application

All trans-Retinal has been used:
  • in optogenetic experiments
  • in electrophysiological experiment
  • to study the effect of AKR1B10 (aldo-keto reductase (AKR) superfamily member) on the conversion of retinal to retinol in the airway epithelium
  • in decidual transformation of human endometrial stromal cells

Biochem/physiol Actions

All-trans retinal is converted to retinoic acid in vivo by the action of retinal dehydrogenase. Retinoic acid is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) that act as transcription factors to regulate the growth and differentiation of normal and malignant cells. Retinal isomers are also chromophores that bind to opsins, a family of G-protein-linked transmembrane proteins, to form photosensitive receptors in visual and nonvisual systems. All-trans retinal is a potent photosensitizer.

Packaging

Sealed ampule.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Optogenetic and pharmacologic dissection of feedforward inhibition in Drosophila motion vision
Mauss AS, et al.
The Journal of Neuroscience, 34(6), 2254-2263 (2014)
Shiho Kawamura et al.
Structure (London, England : 1993), 21(3), 426-437 (2013-02-26)
Rhodopsin, the photoreceptor pigment of the retina, initiates vision upon photon capture by its covalently linked chromophore 11-cis-retinal. In the absence of light, the chromophore serves as an inverse agonist locking the receptor in the inactive dark state. In the
Ye Zhang et al.
Nature communications, 10(1), 1560-1560 (2019-04-07)
In the Drosophila antenna, different subtypes of olfactory receptor neurons (ORNs) housed in the same sensory hair (sensillum) can inhibit each other non-synaptically. However, the mechanisms underlying this underexplored form of lateral inhibition remain unclear. Here we use recordings from
Alexandria H Jaeger et al.
eLife, 7 (2018-10-12)
Each taste modality is generally encoded by a single, molecularly defined, population of sensory cells. However, salt stimulates multiple taste pathways in mammals and insects, suggesting a more complex code for salt taste. Here, we examine salt coding in Drosophila.
Lieuwe Biewenga et al.
ACS omega, 5(5), 2397-2405 (2020-02-18)
The efficient engineering of iminium biocatalysis has drawn considerable attention, with many applications in pharmaceutical synthesis. Here, we report a tailor-made iminium-activated colorimetric "turn-on" probe, specifically designed as a prescreening tool to facilitate engineering of iminium biocatalysis. Upon complexation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service