Skip to Content
Merck
All Photos(3)

Documents

H6278

Sigma-Aldrich

4-Hydroxytamoxifen

≥98% (HPLC), powder, Tamoxifen metabolite

Synonym(s):

4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, 4-OHT, cis/trans-4-Hydroxytamoxifen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29NO2
CAS Number:
Molecular Weight:
387.51
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

product name

4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

solubility

methanol: 10 mg/mL
ethanol: 20 mg/mL (with heating)

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

CC\C(c1ccccc1)=C(/c2ccc(O)cc2)c3ccc(OCCN(C)C)cc3.CC\C(c4ccccc4)=C(\c5ccc(O)cc5)c6ccc(OCCN(C)C)cc6

InChI

1S/2C26H29NO2/c2*1-4-25(20-8-6-5-7-9-20)26(21-10-14-23(28)15-11-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h2*5-17,28H,4,18-19H2,1-3H3/b26-25+;26-25-

InChI key

ZJLDABGSDWXVGE-BDSXMVAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxytamoxifen (4-OHT), a first-generation selective estrogen receptor modulator (SERM), acts as an antagonist in breast cancer cells but exhibits estrogen-like effects in the uterus and bone. It is an active metabolite of the pro-drug tamoxifen (TAM), functioning as an estrogen receptor antagonist. 4-OHT binds to estrogen receptors to inhibit estradiol-induced cell proliferation without inducing cytotoxicity. It is a metabolite of the antiestrogen, tamoxifen and exhibits a higher affinity than tamoxifen and its other metabolites for binding to estrogen receptors, making it 50 to 100 times more potent in inhibiting cell proliferation in normal human breast cells and breast cancer cell lines in culture.

4-OHT effectively inhibited cell growth in the absence of estrogen when cell proliferation was stimulated by insulin or epidermal growth factor. 4-OHT inhibits lipid peroxidation within cell membranes and shows peroxyl radical scavenging activity.

Application

4-Hydroxytamoxifen has been used:
  • to induce the recombination of small intestinal organoids.
  • to study its impact on the ability of human peripheral blood mononuclear cells (PMNCs) to form hematopoietic colonies.
  • to induce overexpression of MYCN in the neuroblastoma cell line to determine how the elevated MYCN expression influences the sensitivity of neuroblastoma cells to BIRC5/survivin inhibitor, YM155-induced apoptosis.

Biochem/physiol Actions

Metabolite of the chemotherapeutic drug tamoxifen, exhibiting more potent estrogen agonist/antagonist activity than the parent drug. Also active as intra-membranous inhibitor of lipid peroxidation.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-Hydroxytamoxifen is an isoform-specific inhibitor of orphan estrogen-receptor-related (ERR) nuclear receptors beta and gamma
Tremblay GB, et al.
Endocrinology, 142, 4572-4575 (2001)
YM155 inhibits neuroblastoma growth through degradation of MYCN: A new role as a USP7 inhibitor
Li X, et al.
Journal of Separation Science, 181, 106343-106343 (2023)
Tamoxifen and hydroxytamoxifen as intramembraneous inhibitors of lipid peroxidation. Evidence for peroxyl radical scavenging activity
Cust'odio JBA, et al.
Biochemical Pharmacology, 47, 1989-1998 (1994)
4-Hydroxytamoxifen is an isoform-specific inhibitor of orphan estrogen-receptor-related (ERR) nuclear receptors \beta and \gamma
Tremblay, Gilles B and Bergeron, Denis and Giguere, Vincent
Journal of Separation Science, 142, 4572-4575 (2001)
Tamoxifen for the treatment of myeloproliferative neoplasms: A Phase II clinical trial and exploratory analysis
Fang Z, et al.
Nature Communications, 14(1), 7725-7725 (2023)

Articles

We presents an article on Autophagy in Cancer Promotes Therapeutic Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service