Skip to Content
Merck
All Photos(1)

Key Documents

579000

Sigma-Aldrich

Tamoxifen Citrate

Tamoxifen is a potent synthetic anti-estrogenic agent.

Synonym(s):

Tamoxifen Citrate, Estrogen Receptor Signaling Regulator I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29NO · xC6H8O7
CAS Number:
Molecular Weight:
371.51 (salt-free basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

description

Merck USA index - 14, 9048

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

ethanol: 5 mg/mL
methanol: 5 mg/mL
water: sparingly soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(CCOc1ccc(cc1)\C(=C(\CC)/c3ccccc3)\c2ccccc2)(C)C.[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].[H+].[H+].[H+]

InChI

1S/C26H29NO.C6H8O7/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-18H,4,19-20H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;

InChI key

FQZYTYWMLGAPFJ-OQKDUQJOSA-N

General description

A potent synthetic anti-estrogen. A cell-permeable and reversible inhibitor of protein kinase C (IC50 = 10 µM). Induces apoptosis in human malignant glioma cell lines.
A potent synthetic anti-estrogenic agent. A cell-permeable and reversible inhibitor of protein kinase C (IC50 = 10 µM). Induces apoptosis in human malignant glioma cell lines and inhibits prostate cancer cell growth by induction of p21 protein.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product does not compete with ATP.
Reversible: yes
Target IC50: 10 µM against protein kinase C

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rohlff, C., et al. 1998. Prostate 37, 51.
Couldwell, W.J., et al. 1994. FEBS Lett.345, 43.
Baltuch, G.H., et al. 1993. Neurosurgery33, 495.
Powis, G. 1991. Trends Pharmacol. Sci.12, 188.
Issandou, M., et al. 1990. Cancer Res.50, 5845.
O’Brian, C.A., et al. 1985. Cancer Res.45, 2462.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viney Kumar et al.
3 Biotech, 13(6), 193-193 (2023-05-19)
Converging evidences identifies that microRNA-21 (miR-21) is responsible for drug resistance in breast cancer. This study aims to evaluate the miR-21-modulatory potential of a hybrid compound, pterostilbene-isothiocyanate (PTER-ITC), in tamoxifen-resistant MCF-7 (TR/MCF-7) and 5-fluorouracil-resistant MDA-MB 231 (5-FUR/MDA-MB 231) breast cancer
Flavio C Lombardo et al.
ACS infectious diseases, 7(7), 1894-1900 (2020-10-28)
Schistosomiasis is a major neglected tropical disease with more than 200 million infections annually. Despite only one drug, praziquantel, being available, the drug pipeline against schistosomiasis is empty, and drug screening tools have limitations. We evaluated the potential of human
Aarthi Sekar et al.
eNeuro, 8(6) (2021-11-03)
Tracking and quantifying the abundance and location of cells in the developing brain is essential in neuroscience research, enabling a greater understanding of mechanisms underlying nervous system morphogenesis. Widely used experimental methods to quantify cells labeled with fluorescent markers, such
Feng Yue et al.
Cell reports, 38(3), 110267-110267 (2022-01-20)
The lipid droplet (LD) is a central hub for fatty acid metabolism in cells. Here we define the dynamics and explore the role of LDs in skeletal muscle satellite cells (SCs), a stem cell population responsible for muscle regeneration. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service