Skip to Content
Merck
All Photos(1)

Key Documents

SML0919

Sigma-Aldrich

ML221

≥98% (HPLC)

Synonym(s):

5-[(4-Nitrobenzoyl)oxy]-2-[(2-pyrimidinylthio)methyl]-4H-pyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N3O6S
CAS Number:
Molecular Weight:
385.35
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

S(Cc2[o]cc([c](c2)=O)OC(=O)c3ccc(cc3)[N+](=O)[O-])c1ncccn1

InChI

1S/C17H11N3O6S/c21-14-8-13(10-27-17-18-6-1-7-19-17)25-9-15(14)26-16(22)11-2-4-12(5-3-11)20(23)24/h1-9H,10H2

InChI key

UASIRTUMPRQVFY-UHFFFAOYSA-N

Biochem/physiol Actions

ML221 is an antagonist of the apelin receptor (also known as APJ).
ML221 is an antagonist of the apelin receptor (also known as APJ). ML221 blocks Apelin-13 mediated receptor activation (recruitment of β-arrestin) with an IC50 of 1.75 μM.
ML221 is capable of suppressing the cultured-endothelial cell proliferation. In oxygen-induced retinopathy (OIR) mice model, ML221 can block the pathological angiogenesis and increase the recovery of normal vessels into the ischemic regions in the retina. It can also inhibit the formation of neovascular tufts.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An apelin receptor antagonist prevents pathological retinal angiogenesis with ischemic retinopathy in mice
Ishimaru Y, et al.
Scientific reports, 7(15062), 1-9 (2017)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service