Skip to Content
Merck
All Photos(3)

Key Documents

G7252

Sigma-Aldrich

Guanosine 5′-diphosphate tris salt from Saccharomyces cerevisiae

Type VI, ≥92.5%

Synonym(s):

GDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O11P2
CAS Number:
Molecular Weight:
443.20
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

type

Type VI

Quality Level

Assay

≥92.5%

shipped in

dry ice

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H15N5O11P2.C4H11NO3/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(25-9)1-24-28(22,23)26-27(19,20)21;5-4(1-6,2-7)3-8/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H2,19,20,21)(H3,11,13,14,18);6-8H,1-3,5H2/t3-,5-,6-,9-;/m1./s1

InChI key

YWMHXINREUQYRO-GWTDSMLYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Guanosine 5′-diphosphate (GDP) is the diphosphate nucleoside of the purine guanine. GDP and GTP are involved in and may be used to study cell signaling via G-coupled protein receptors (GCPR) and guanine nucleotide exchange factors (GEF) and a variety of GTPases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola M Martucci et al.
Biochimie, 94(2), 503-509 (2011-09-20)
The effect of pulvomycin on the biochemical and fluorescence spectroscopic properties of the archaeal elongation factor 1α from Sulfolobus solfataricus (SsEF-1α), the functional analog of eubacterial EF-Tu, was investigated. The antibiotic was able to reduce in vitro the rate of protein
Jason Lowery et al.
The Journal of biological chemistry, 286(42), 36898-36906 (2011-08-11)
ADP-ribosylation factors (ARFs) and their activating guanine nucleotide exchange factors (GEFs) play key roles in membrane traffic and signaling. All ARF GEFs share a ∼200-residue Sec7 domain (Sec7d) that alone catalyzes the GDP to GTP exchange that activates ARF. We
Antonio Castro-Castro et al.
The EMBO journal, 30(19), 3913-3927 (2011-08-30)
The activation of the Rac1 GTPase during cell signalling entails its translocation from the cytosol to membranes, release from sequestering Rho GDP dissociation inhibitors (RhoGDI), and GDP/GTP exchange. In addition to those steps, we show here that optimal Rac1 activation
Sara J Wright et al.
Genetics, 189(1), 165-176 (2011-07-14)
Heterotrimeric (αβγ) G proteins are crucial components of eukaryotic signal transduction pathways. G-protein-coupled receptors (GPCRs) act as guanine nucleotide exchange factors (GEFs) for Gα subunits. Recently, facilitated GDP/GTP exchange by non-GPCR GEFs, such as RIC8, has emerged as an important
Rachel Cooley et al.
Wellcome open research, 5, 20-20 (2020-06-27)
Targeting the interaction of proteins with weak binding affinities or low solubility represents a particular challenge for drug screening. The NanoLuc â ® Binary Technology (NanoBiT â ®) was originally developed to detect protein-protein interactions in live mammalian cells. Here we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service