Skip to Content
Merck

Skip To

C7252

D-(+)-Cellobiose

≥98% (HPLC)

Synonym(s):

β-D-Glc-(1→4)-D-Glc, 4-O-β-D-Glucopyranosyl-D-glucose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352201
NACRES:
NA.21
PubChem Substance ID:
EC Number:
208-436-5
Beilstein/REAXYS Number:
93795
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant

Quality Level

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 34±1, c = 10% (w/v) in water

technique(s)

HPLC: suitable

color

beige

mp

239 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h1,4-12,14-21H,2-3H2/t4-,5+,6+,7+,8+,9-,10+,11+,12-/m0/s1

InChI key

DKXNBNKWCZZMJT-WELRSGGNSA-N

General description

Cellobiose, a disaccharide, is made up of two d-glucose molecules linked by a β-1,4-glycosidic bond. This reducing sugar can mutarotate and is produced by hydrolysis of cellulose.

Application

D-(+)-Cellobiose has been used:
  • as a component in test sugar solution for cellobiose-mannitol permeability test
  • in the preparation of lyophilization solutions to study its ability to protect lyophilized β-galactosidase from enzymatic activity loss and secondary structure changes during storage
  • as a fermentation/growth substrate to grow Clostridium thermocellum to study its impacts on the qualitative and quantitative changes in cellulosome composition

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2215091852219458
technique(s)

HPLC: suitable

technique(s)

-

technique(s)

-

technique(s)

-

biological source

plant

biological source

-

biological source

-

biological source

-

assay

≥98% (HPLC)

assay

≥99.0% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

solubility

water: 50 mg/mL, clear, colorless

solubility

-

solubility

-

solubility

water: 5 mg/mL

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

form

powder

form

powder

form

-

form

crystalline powder


Still not finding the right product?

Explore all of our products under D-(+)-Cellobiose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Mireia Lopez-Siles et al.
Applied and environmental microbiology, 78(2), 420-428 (2011-11-22)
Faecalibacterium prausnitzii is one of the most abundant commensal bacteria in the healthy human large intestine, but information on genetic diversity and substrate utilization is limited. Here, we examine the phylogeny, phenotypic characteristics, and influence of gut environmental factors on
Shuen Hon et al.
Metabolic engineering communications, 3, 120-129 (2016-04-22)
Clostridium thermocellum is a promising candidate for ethanol production from cellulosic biomass, but requires metabolic engineering to improve ethanol yield. A key gene in the ethanol production pathway is the bifunctional aldehyde and alcohol dehydrogenase, adhE. To explore the effects
R Carratù et al.
Journal of pediatric gastroenterology and nutrition, 28(3), 264-269 (1999-03-06)
Intestinal permeability has seldom been investigated in diabetes mellitus, even though patients frequently report gastrointestinal symptoms, and it has recently been shown that the prevalence of celiac disease associated with diabetes mellitus is higher than expected. Intestinal permeability to cellobiose



Global Trade Item Number

SKUGTIN
C7252-500G04061835543526
C7252-10MG04061833512036
C7252-1KG04061833512043
C7252-25G04061835543519
C7252-100G04061835543502

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters