Skip to Content
Merck
All Photos(3)

Key Documents

G6046

Sigma-Aldrich

Anti-G3BP antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-G3BP1, Anti-GAP SH3 domain-binding protein, Anti-HDH-VIII, Anti-Ras-GTPase-activating protein SH3-domain binding protein 1, Anti-human DNA helicase VIII

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~68 kDa

species reactivity

human, mouse (predicted), rat (predicted)

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2-5 μg/mL using human HeLa cells
western blot: 0.1-0.2 μg/mL using whole extract of human U-2-OS cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G3BP1(10146)
mouse ... G3bp1(27041)
rat ... G3bp1(171092)

General description

G3BP is a RNA-binding protein that contains a RNA binding domain, the RRM-type domain, at the carboxyl-terminal. The N-terminal domain is homologous to nuclear transporter factor 2 (NTF2) and the central domain is rich in acidic residues. The RRM domain is involved in the binding of G3BP to specific RNA sequences. This helps G3BP to function as a CA dinucleotide-specific endoribonuclease. G3BP′s subcellular localization and protein-protein interactions are mediated by the phosphorylation of G3BP at Ser149, near NTF2-like domain. G3BP is found in abundance in many kinds of malignant tumors such as lung, colon, gastric, and breast cancer.

Application

Anti-G3BP antibody produced in rabbit has been used in co-immunoprecipitation assay.
Anti-G3BP antibody produced in rabbit has been used in
  • immunofluorescence analysis
  • immunostaining
  • immunoblotting
  • immunoprecipitation

Biochem/physiol Actions

G3BP (Ras-GTPase-activating protein SH3 domain binding protein 1) is a endoribonuclease that is phosphorylation-dependent and specific to single strand. It cleaves between cytosine and adenine (CA). It binds to the SH3 domain of RasGAP. It forms a link between RasGAP-mediated signaling pathway and RNA turnover. G3BP is required for the assembly of stress granules (SGs), which are involved in the regulation of mRNA metabolism during stress. It may be used as a cytoplasmic SG marker.
Ras-GAP SH3 domain binding protein (G3BP) is overexpressed in many kinds of malignant tumors such as breast, colon, lung and gastric breast cancer. G3BP expression is correlated with lymph node metastasis in breast cancer.

Target description

G3BP encodes one of the DNA-unwinding enzymes which prefers partially unwound 3′-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

In immunoblotting assays, it is highly recommended to dilute the antibody in 5% NFDM (w/v).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelke Jan Fros et al.
eLife, 6 (2017-09-30)
Most vertebrate and plant RNA and small DNA viruses suppress genomic CpG and UpA dinucleotide frequencies, apparently mimicking host mRNA composition. Artificially increasing CpG/UpA dinucleotides attenuates viruses through an entirely unknown mechanism. Using the echovirus 7 (E7) model in several
Probing protein interactions in living mammalian cells on a microtubule bench
Boca M, et al.
Scientific reports, 5, 17304-17304 (2015)
Takuya Sueyoshi et al.
Journal of immunology (Baltimore, Md. : 1950), 200(11), 3814-3824 (2018-04-22)
Retinoic acid-inducible gene I (RIG-I)-like receptors (RLRs), RIG-I, and melanoma differentiation-associated gene 5 (MDA5) play a critical role in inducing antiviral innate immune responses by activating IFN regulatory factor 3 (IRF3) and NF-κB, which regulates the transcription of type I
Giel P Göertz et al.
PLoS neglected tropical diseases, 12(11), e0006958-e0006958 (2018-11-10)
Chikungunya virus (CHIKV) is a re-emerging arthropod-borne (arbo)virus that causes chikungunya fever in humans and is predominantly transmitted by Aedes aegypti mosquitoes. The CHIKV replication machinery consists of four non-structural proteins (nsP1-4) that additionally require the presence of a number
Yan Zheng et al.
Journal of virology, 87(17), 9579-9589 (2013-06-21)
Alphaviruses are enveloped viruses with highly organized structures. The nucleocapsid (NC) core contains a capsid protein lattice enclosing the plus-sense RNA genome, and it is surrounded by a lipid bilayer containing a lattice of the E1 and E2 envelope glycoproteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service