Skip to Content

936B

Emerald: Antibody Diluent

PBS, <1% BSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
12352203
Technique(s):
immunohistochemistry: suitable
Species reactivity:
-
Application:
IHC
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

[For In Vitro Diagnostic Use in Select Regions]

form

buffered aqueous solution (green)

packaging

pkg of 1 L ready to use (936B-09), pkg of 250 mL ready to use (936B-08), pkg of 50 mL ready to use (936B-03)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Emerald: Antibody Diluent is an appropriately buffered solution for the dilution of both polyclonal and monoclonal antibodies as well as for the preparation of negative control reagents in IHC. It is designed to minimize the non-specific reaction that may be caused by the antibody and encourage specific antigen-antibody binding. Emerald: Antibody Diluent can also be used to stabilize diluted antibodies when stored at 2-8°C.

Physical form

Solution containing PBS buffer, pH 7.3-7.7, with 1% BSA and <0.1% sodium azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library