Skip to Content
Merck

Skip To

F9261

Flunitrazepam

Synonym(s):

5-(2-Fluorophenyl)-1-methyl-7-nitro-3H-1,4-benzodiazepin-2(1H)-one, Ro 5-4200

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H12FN3O3
CAS Number:
Molecular Weight:
313.28
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
216-597-8
MDL number:
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1 mg/mL, DMF: soluble, ethanol: soluble

originator

Roche

SMILES string

CN1C(=O)CN=C(c2ccccc2F)c3cc(ccc13)[N+]([O-])=O

InChI

1S/C16H12FN3O3/c1-19-14-7-6-10(20(22)23)8-12(14)16(18-9-15(19)21)11-4-2-3-5-13(11)17/h2-8H,9H2,1H3

InChI key

PPTYJKAXVCCBDU-UHFFFAOYSA-N

Biochem/physiol Actions

Hypnotic; anxiolytic; ligand for the GABAA receptor benzodiazepine modulatory site.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D0899P3654L1764
drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1 mg/mL, ethanol: soluble, DMF: soluble

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1.6 mg/mL, DMF: soluble, H2O: slightly soluble, ethanol: soluble

solubility

-

solubility

-

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)
rat ... Gabra2(29706)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)
rat ... Gabra1(29705), Gabra2(29706)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

Gene Information

human ... GABRA1(2554), GABRA2(2555), GABRA3(2556), GABRA4(2557), GABRA5(2558), GABRA6(2559), GABRB1(2560), GABRB2(2561), GABRB3(2562), GABRD(2563), GABRE(2564), GABRG1(2565), GABRG2(2566), GABRG3(2567), GABRP(2568), GABRQ(55879)

originator

Roche

originator

Roche

originator

-

originator

Baxter


Still not finding the right product?

Explore all of our products under Flunitrazepam


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


R M McKernan et al.
Molecular pharmacology, 54(1), 33-43 (1998-07-11)
Photoincorporation of ligands into the benzodiazepine site of native gamma-aminobutyric acidA (GABAA) receptors provides useful information about the nature of the benzodiazepine (BZ) binding site. Photoincorporation of flunitrazepam into a single population of GABAA receptors, recombinant human alpha1beta3gamma2, was investigated
Danni Harris et al.
Journal of medicinal chemistry, 51(13), 3788-3803 (2008-06-10)
Ligands that bind to the benzodiazepine binding site on the GABA A receptor can attenuate or potentiate cognition. To investigate this property, the chemical determinants favoring selective binding or selective activation of the alpha5beta2gamma2 and alpha1beta2gamma2 GABA A receptor isoforms
Mari Takeuchi et al.
Annals of surgical oncology, 19(12), 3963-3970 (2012-06-16)
Postoperative delirium is a common complication after major surgery and is characterized by acute confusion with fluctuating consciousness. The aim of this study was to investigate the incidence and risk factors of postoperative delirium in patients with esophageal cancer. We



Global Trade Item Number

SKUGTIN
F9261-50MG04061833621486
F9261-100MG04061832556574

Questions

Reviews

No rating value

Active Filters