Skip to Content
Merck
All Photos(1)

Key Documents

H9149

Sigma-Aldrich

α-Hydroxyalprazolam

powder, ≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN4O
CAS Number:
Molecular Weight:
324.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

drug control

Home Office Schedule 4.1; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to off-white

solubility

methylene chloride: ~20 mg/mL, clear, colorless to faintly yellow

SMILES string

OCc1nnc2CN=C(c3ccccc3)c4cc(Cl)ccc4-n12

InChI

1S/C17H13ClN4O/c18-12-6-7-14-13(8-12)17(11-4-2-1-3-5-11)19-9-15-20-21-16(10-23)22(14)15/h1-8,23H,9-10H2

InChI key

ZURUZYHEEMDQBU-UHFFFAOYSA-N

Caution

store dessicated and protected from light

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Smith et al.
Psychopharmacology, 93(1), 105-112 (1987-01-01)
The relationships between alprazolam and metabolite concentrations and CNS effects were determined in a double-blind placebo controlled four-way crossover trial in 16 normal male volunteers. Active drug treatments consisted of 4-day regimens of 4 mg alprazolam PO daily as 2
V D Schmith et al.
Journal of clinical pharmacology, 31(6), 571-579 (1991-06-01)
In a double-blind, randomized, placebo-controlled study, the pharmacokinetics of alprazolam and its active metabolite, alpha-hydroxyalprazolam, were determined in 12 normal subjects and 12 dialysis patients [7 hemodialysis (HD) patients and 5 continuous ambulatory peritoneal dialysis (CAPD) patients]. Blood samples were
K M Höld et al.
Journal of mass spectrometry : JMS, 31(9), 1033-1038 (1996-09-01)
A sensitive and specific method was developed for the determination of alprazolam and its major metabolite alpha-hydroxyalprazolam in plasma. After the addition of deuterium-labeled internal standards, plasma samples were buffered to pH 9 with 1 ml of saturated sodium borate
D J Crouch et al.
Journal of analytical toxicology, 23(6), 479-485 (1999-10-12)
A sensitive and specific electrospray ionization high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS-MS) method has been developed for the quantitative determination of alprazolam (AL) and alpha-hydroxyalprazolam (OH-AL) in plasma. After the addition of deuterium labeled internal standards of AL and OH-AL
Annika Allqvist et al.
European journal of clinical pharmacology, 63(2), 173-179 (2007-01-04)
The antifungal drug ketoconazole (KTZ) is known as an inhibitor of, especially, the CYP3A subfamily, which catalyzes the metabolism of a large variety of drugs. Interactions between KTZ and CYP3A substrates have been reported both in vivo and in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service