F4004
β-Fluoropyruvic acid sodium salt monohydrate
≥98%
Synonym(s):
sodium 3-fluoro-2-oxopropanoate hydrate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
FCH2COCO2Na · H2O
CAS Number:
Molecular Weight:
146.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥98%
storage temp.
−20°C
SMILES string
FCC(C([O-])=O)=O.O.[Na+]
InChI
1S/C3H3FO3.Na.H2O/c4-1-2(5)3(6)7;;/h1H2,(H,6,7);;1H2/q;+1;/p-1
InChI key
HDSZBLZMLDQKRW-UHFFFAOYSA-M
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
E B Borthwick et al.
The Biochemical journal, 305 ( Pt 2), 521-524 (1995-01-15)
Simplified procedures for assaying and purifying dihydrodipicolinate synthase (EC 4.2.1.52), the first enzyme of the lysine biosynthetic pathway, have been developed and electrospray ionization m.s. has been used to observe the imine intermediate with pyruvate and to investigate the reaction
Fluoropyruvate: a potent inhibitor of the bacterial and the mammalian pyruvate dehydrogenase complex.
H Bisswanger
Biochemical and biophysical research communications, 95(2), 513-519 (1980-07-31)
A Balme et al.
Protein science : a publication of the Protein Society, 3(1), 109-117 (1994-01-01)
The family of FMN-dependent, alpha-hydroxy acid-oxidizing enzymes catalyzes substrate dehydrogenation by a mechanism the first step of which is abstraction of the substrate alpha-proton (so-called carbanion mechanism). For flavocytochrome b2 and lactate oxidase, it was shown that once on the
Lisa M Eubanks et al.
Biochemistry, 42(4), 1140-1149 (2003-01-29)
1-Deoxy-d-xylulose 5-phosphate synthase (DXP synthase) catalyzes the thiamine diphosphate (TPP)-dependent condensation of pyruvate and d-glyceraldehyde 3-phosphate (GAP) to yield DXP in the first step of the methylerythritol phosphate pathway for isoprenoid biosynthesis. Steady-state kinetic constants for DXP synthase calculated from
L G Korotchkina et al.
Archives of biochemistry and biophysics, 369(2), 277-287 (1999-09-16)
Pyruvate dehydrogenase (E1), a heterotetramer (alpha(2)beta(2)), is the first catalytic component of the mammalian pyruvate dehydrogenase complex (PDC). To investigate the roles of cysteine-62 of E1alpha (alphaC62) and tryptophan-135 of E1beta (betaW135) (identified previously as active site residues using chemical
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service