Skip to Content
Merck
All Photos(3)

Key Documents

S2889

Sigma-Aldrich

Sodium acetate

anhydrous, for molecular biology, ≥99%

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021906
PubChem Substance ID:
NACRES:
NA.25

grade

Molecular Biology
anhydrous
for molecular biology

Quality Level

Assay

≥99%

form

solid

autoignition temp.

1112 °F

pH

8.5-9.9 (25 °C, 246 g/L)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL

foreign activity

DNase, RNase, protease, none detected

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Evaluating acetate as a renoprotective agent following kidney ischemia in a porcine model.: This study explores the use of sodium acetate as a renoprotective agent to mitigate kidney damage following ischemia, highlighting its potential therapeutic applications in renal protection (Kominsky HD et al., 2024).

  • Chitosan-PVA-PVP/nano-clay composite: a promising tool for controlled drug delivery.: Sodium acetate plays a crucial role in the synthesis of chitosan-based nanocomposites for controlled drug delivery systems, demonstrating its significance in pharmaceutical applications (Ali M et al., 2024).

  • Direct Affinity Ligand Immobilization onto Bare Iron Oxide Nanoparticles Enables Efficient Magnetic Separation of Antibodies.: Sodium acetate is utilized in the immobilization process of affinity ligands onto nanoparticles for efficient magnetic separation, highlighting its role in biochemical separations and diagnostics (Zimmermann I et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulivan Jouanneau et al.
Environmental science & technology, 46(21), 11979-11987 (2012-09-20)
In this study, we compared two bacterial biosensors designed for the environmental monitoring of metals: Lumisens III and Lumisens IV. These two biosensors are based on the same bacterial sensors (inducible or constitutive bacterial strains) but with a different conservation
Jamie Morrison et al.
The American journal of pathology, 166(6), 1701-1710 (2005-05-28)
Duchenne muscular dystrophy was initially described as a myosclerosis because of the conspicuous progression of interstitial fibrosis. Using the mdx mouse homologue, we have shown previously that the accumulation of intramuscular collagen is profoundly influenced by the presence or absence
Mari Endo et al.
Journal of gastroenterology, 49(6), 1026-1039 (2013-07-13)
Daikenchuto (DKT), a gastrointestinal prokinetic Japanese herbal medicine, is prescribed for patients with postoperative ileus (POI) and adhesive bowel obstruction following abdominal surgery. Several mechanisms for the amelioration of POI by DKT have been suggested; however, it has remained unclear
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils
Amanda P De Souza et al.
Journal of experimental botany, 66(14), 4351-4365 (2015-04-25)
The production of bioenergy from grasses has been developing quickly during the last decade, with Miscanthus being among the most important choices for production of bioethanol. However, one of the key barriers to producing bioethanol is the lack of information

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service