Skip to Content
Merck
All Photos(3)

Documents

117714

Sigma-Aldrich

3-Methoxybenzoic acid

ReagentPlus®, 99%

Synonym(s):

m-Anisic acid, m-Methylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
508838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

bp

170-172 °C/10 mmHg (lit.)

mp

105-107 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear, colorless to faintly yellow

SMILES string

COc1cccc(c1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

XHQZJYCNDZAGLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxybenzoic acid is an important intermediate in the synthesis of natural products.

Application

3-Methoxybenzoic acid was used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III). It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium borohydride reduction of aromatic carboxylic acids via methyl esters.
Saeed A and Ashraf Z.
Journal of Chemical Sciences (Bangalore), 118(5), 419-423 (2006)
Synthesis, characterization and thermal behaviour of solid-state compounds of europium (III) and gadolinium (III) 3-methoxybenzoate.
Dametto PR, et al.
Journal of Thermal Analysis and Calorimetry, 97(2), 765-768 (2009)
A S Waldman et al.
Nucleic acids research, 19(21), 5943-5947 (1991-11-11)
We determined the effect of 3-methoxybenzamide (3-MB), a competitive inhibitor of poly(ADP-ribose) polymerase (E.C. 2.4.2.30), on intrachromosomal homologous recombination in mouse Ltk- cells. We used a cell line that contained in its genome two defective Herpes thymidine kinase (tk) genes
D H Lee et al.
Xenobiotica; the fate of foreign compounds in biological systems, 29(9), 909-916 (1999-11-05)
1. 2-(Allylthio)pyrazine (2-AP) has been demonstrated to protect the liver against toxicants by inhibiting CYP2E1 activity. Since 2-mercaptopyrazine (2-MP) is presumed to be a metabolite of 2-AP, the experiments were performed to determine whether rat liver microsomal and/or cytosolic preparations
Thi-Huu Nguyen et al.
Organic letters, 7(12), 2445-2448 (2005-06-04)
[reaction: see text] If employed in THF at 0 degrees C, LTMP metalates meta-anisic acid at the doubly activated position. In contrast, n-BuLi/t-BuOK deprotonates position C-4 preferentially at low temperature. Functionalization at C-6 requires protection of the C-2 site beforehand.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service