Skip to Content
Merck
All Photos(1)

Key Documents

F0301

Sigma-Aldrich

Monoclonal Anti-CD56-FITC antibody produced in mouse

clone C5.9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-CD56, Anti-NKH-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C5.9, monoclonal

form

buffered aqueous solution

mol wt

antigen 150 kDa

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NCAM1(4684)

Looking for similar products? Visit Product Comparison Guide

General description

Prepared by conjugation of fluorescein isothiocyanate isomer 1 (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Immunogen

A cloned human NK cell line termed JT3

Application

Monoclonal Anti-CD56-FITC antibody produced in mouse is suitable for fluorescence microscopy. It was also used for flow cytometric analysis.

Biochem/physiol Actions

Neural Cell Adhesion Molecule (NCAM or the cluster of differentiation CD56) is a homophilic binding glycoprotein. It is expressed on the surface of neurons, glia, skeletal muscle and natural killer cells. CD56 can also be expressed in normal immature granulocytes at different expression levels in regenerative bone marrow. It is associated with cardiac genes induced by metabolic stress and its expression is associated with worsening hemodynamic parameters and major metabolic genes. CD56 is involved in left ventricular remodeling and can be implicated in heart failure. It is is developmentally regulated and may have an important role in cardiac development.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 0.2% gelatin and 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuo Muroi et al.
Journal of clinical and experimental hematopathology : JCEH, 53(3), 247-250 (2013-12-27)
Bone marrow mononuclear cells from 93 patients with hematological malignancies after allogeneic hematopoietic stem cell transplantation (AHSCT) were analyzed using flow cytometry (FCM). The disease was acute myeloblastic leukemia (50 patients), acute lymphoblastic leukemia, and others. Conditioning was myeloablative (80
Branka Petricevic et al.
Journal of translational medicine, 11, 307-307 (2013-12-18)
Monoclonal antibodies (mAb), such as trastuzumab are a valuable addition to breast cancer therapy. Data obtained from neoadjuvant settings revealed that antibody-dependent cell-mediated cytotoxicity (ADCC) is a major mechanism of action for the mAb trastuzumab. Conflicting results still call into
Kazuya Nagao et al.
Circulation. Heart failure, 7(2), 351-358 (2013-12-25)
Recently, we screened for cardiac genes induced by metabolic stress and identified neural cell adhesion molecule (NCAM) as a candidate. This study aimed to clarify the expression pattern of NCAM in human cardiomyopathy. A total of 64 cardiac tissue samples
A A Reyes et al.
Molecular and cellular biology, 11(3), 1654-1661 (1991-03-01)
The major membrane-associated or transmembrane isoforms of the neural cell adhesion molecule (NCAM) are generated by alternative splicing at the 3' end of the mRNA. Further diversity in NCAM structure is observed in the extracellular region of the polypeptide, where
Takaki Asano et al.
Frontiers in immunology, 9, 568-568 (2018-04-21)
Activated PI3Kδ syndrome (APDS) is a primary immunodeficiency characterized by recurrent respiratory tract infections, lymphoproliferation, and defective IgG production. Heterozygous mutations in PIK3CD, PIK3R1, or PTEN, which are related to the hyperactive phosphoinositide 3-kinase (PI3K) signaling, were recently presented to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service