Skip to Content
Merck
All Photos(1)

Key Documents

A2327

Sigma-Aldrich

Adrenomedullin 52 human

≥90% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C264H406N80O77S3
CAS Number:
Molecular Weight:
6028.73
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ADM(133)

Amino Acid Sequence

Tyr-Arg-Gln-Ser-Met-Asn-Asn-Phe-Gln-Gly-Leu-Arg-Ser-Phe-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Asn-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2

General description

Adrenomedullin (ADM) human comprises of 52 amino acids and has amine group in the C-terminal tyrosine residue. It also has a N-terminal ring structure and belongs to calcitonin peptide family. ADM has vasodilatory function and is mapped to human chromosome 11p15.4. Adrenomedullin is highly expressed in endothelial cells. Adrenomedullin controls a majority of cellular processes using cyclic adenosine monophosphate (cAMP) as a messenger. It interacts with calcitonin receptor-like receptor (CRLR) and receptor activity modifying protein 2 (RAMP2). ADM contributes to invasion activity in brain tumor cell lines. ADM signalling plays a key role in tumor progression, especially in acute myeloid leukemia.

Application

Adrenomedullin 52 human has been used in tube formation assay in human umbilical vein endothelial cells (HUVECs).

Biochem/physiol Actions

Elicits a potent hypotensive effect; plays a key role in regulating endometrial angiogenesis.
Peptide elicits potent hypotensive effect, comparable to calcitonin gene-related peptide. Plays a key role in regulating endometrial angiogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ben S T Wong et al.
Biology of reproduction, 88(2), 34-34 (2012-12-18)
Extravillous cytotrophoblast (EVCT) is responsible for trophoblast invasion, which is an important process during placentation. Dysregulation of the process is associated with a wide range of pregnancy complications. Adrenomedullin (ADM) is a polypeptide expressed most abundantly in first-trimester placentas. We
Xiaoling Yang et al.
Acta biochimica et biophysica Sinica, 45(2), 141-148 (2012-11-24)
Intermedin (IMD) is a member of the calcitonin/calcitonin gene-related peptide (CGRP) family and has similar or more potent cardiovascular actions than adrenomedullin (ADM) and any other CGRP. The aim of the present work is to study the effects of IMD1-53
Transcriptional regulation of adrenomedullin by oncostatin M in human astroglioma cells: Implications for tumor invasion and migration.
Lim SY, et al.
Scientific Reports, 4(4), 6444-6444 (2014)
Philipp Schuetz et al.
Current opinion in infectious diseases, 26(2), 159-167 (2013-02-26)
In patients with community-acquired pneumonia (CAP), blood biomarkers can help to substantially improve individual decisions involving initiation, (de-)intensification, and cessation of antibiotics, and initial risk stratification, site-of-care assignment (outpatient versus ward versus ICU), and discharge. To illustrate these processes, this
Novel peptide antagonists of adrenomedullin and calcitonin gene-related peptide receptors: identification, pharmacological characterization, and interactions with position 74 in receptor activity-modifying protein 1/3.
Robinson, SD et al.
Journal of Pharmacology and Experimental Therapeutics, 331(2), 513-521 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service