Skip to Content

F0522

Monoclonal Anti-CD3−FITC antibody produced in mouse

clone UCHT-1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-CD3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.44
UNSPSC Code:
12352203
Conjugate:
FITC conjugate
Clone:
UCHT-1, monoclonal
Application:
FACS
Citations:
5
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

UCHT-1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 10 μL using 1 × 106 cells

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD3E(916)

General description

CD3e molecule, ε (CD3E) is a polypeptide encoded by the CD3E gene in humans. It resides on chromosome 11. The CD3ε cytoplasmic domain of the T cell receptor binds to the inner leaflet of the plasma membrane (PM) and is accessible during early stages of T cell activation. It is specific for T lymphocytes and is found in Purkinje cells of the cerebellum as well as in gastric parietal cells of humans and other species. Cytoplasmic cyCD3 expression can only be seen during the early phase of NK-cell differentiation.
The CD3 complex (16/20/29/22/25-28 kDa) consists of 5 polypeptide chains associated with each other and with the T cell receptor.

Immunogen

human thymocytes followed by Sezary T cells.

Application

Monoclonal Anti-CD3-FITC antibody produced in mouse is suitable for flow cytometry (at a dilution of 10μL using 1 × 106 cells). It was also used for ADCC Assay.

Biochem/physiol Actions

Recognizes the CD3 complex. The epitope recognized by clone no. UCHT-1 is expressed on the ε-chain of the CD3 antigen/T cell receptor complex. 3rd Workshop: code no. 126, 208 and 471
4th Workshop: paper no. T3.2

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library