Skip to Content
Merck
All Photos(3)

Key Documents

D9760

Sigma-Aldrich

L-(−)-Dithiothreitol

≥95% (titration)

Synonym(s):

(−)-Cleland’s Reagent, (2R,3R)-1,4-Dimercapto-2,3-butanediol, L-DTT, Cleland reagent chiral

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
Beilstein:
2036371
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥95% (titration)

form

powder

color

white

mp

49 - 51 °C ((120 - 124 °F))

solubility

H2O: 50 mg/mL, clear to hazy, colorless

storage temp.

−20°C

SMILES string

O[C@@H](CS)[C@@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m0/s1

InChI key

VHJLVAABSRFDPM-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-DTT [(2R,3R)-1,4-dimercapto-2,3-butanediol; L-dithiothreitol], a chiral bidentate dithiol with two stereogenic centers, may be used in chiroptical response research. L-DDT is proposed as a stereoselective reducing agent for disulfide bridges in complex molecules.[1][2]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hermann Bultmann et al.
Antimicrobial agents and chemotherapy, 51(5), 1596-1607 (2007-01-31)
Previous studies have shown that peptides containing the protein transduction domain (PTD) of the human immunodeficiency virus tat protein (GRKKRRQRRR) were effective inhibitors of herpes simplex virus type 1 (HSV-1) entry (H. Bultmann and C. R. Brandt, J. Biol. Chem.
Stefan Knoppe et al.
Chimia, 67(4), 236-239 (2013-08-24)
Chiral thiolate-protected gold clusters of atomic precision have gained increasing interest in recent years due to their potential use in catalysis, sensing or bioapplications. While the protection of gold clusters with chiral ligands is a rather trivial task, it was
Hiroshi Yao et al.
Langmuir : the ACS journal of surfaces and colloids, 29(21), 6444-6451 (2013-05-03)
We here present a study of cross-linking chemistry of optically inactive monothiol-protected gold clusters by chiral bidentate dithiol with two stereogenic centers, (2R,3R)-1,4-dimercapto-2,3-butanediol (L-dithiothreitol; L-DTT), and explore the impacts of the cross-linking on their chiroptical responses. The pristine protective ligand
Cai Bowen et al.
Cancer research, 79(16), 4124-4134 (2019-06-20)
NKX3.1 is the most commonly deleted gene in prostate cancer and a gatekeeper suppressor. NKX3.1 is a growth suppressor, mediator of apoptosis, inducer of antioxidants, and enhancer of DNA repair. PTEN is a ubiquitous tumor suppressor that is often decreased
Valérie Robin et al.
Molecular therapy. Nucleic acids, 7, 81-89 (2017-06-19)
Spinal muscular atrophy (SMA) is a recessive disease caused by mutations in the SMN1 gene, which encodes the protein survival motor neuron (SMN), whose absence dramatically affects the survival of motor neurons. In humans, the severity of the disease is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service