Skip to Content
Merck
All Photos(1)

Key Documents

C1117

Sigma-Aldrich

Anti-Cy5 antibody, Mouse monoclonal

clone CY5-15, purified from hybridoma cell culture

Synonym(s):

Cy5 Antibody, Cy5 Antibody - Anti-Cy5 antibody, Mouse monoclonal, Monoclonal Anti-Cy5 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CY5-15, monoclonal

form

buffered aqueous solution

packaging

antibody small pack of 25 μL

concentration

~1.5 mg/mL

technique(s)

direct ELISA: suitable
dot blot: 1-2 μg/mL using cell protein extracts labeled with Cy5
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

CyDyes are a family of fluorophores that can be used for labeling proteins, peptides, DNA, RNA, and other biomolecules. These dyes are small, pH insensitive (between pH 3 and pH 10), and soluble in aqueous solution and have tolerance to DMSO. They are more photo-stable than fluorescein, have high molar extinction coefficients, and favorable quantum yields. Due to their physical and chemical properties, they are used (mainly Cy3 and Cy5) in many different biological assays such as: DNA microarrays, protein microarrays, two-dimensional protein analysis (2D gels), fluorescence resonance energy transfer (FRET), and immunocytochemistry.
Mouse monoclonal clone CY5-15 anti-Cy5 antibody recognizes Cy5 and AlexaFluor647 labeled proteins, but not Cy3 labeled proteins.

Specificity

Monoclonal Anti-Cy5 recognizes Cy5 and AlexaFluor647 labeled proteins, but not Cy3 labeled proteins.
The antibody recognizes Cy5 and AlexaFluor 647 labeled proteins, but not Cy3 labeled proteins.

Immunogen

mixture of proteins labeled with Cy5.

Application

Anti-Cy5 antibody, Mouse monoclonal has been used in immunofluorescence assays and western blotting.
Mouse monoclonal clone CY5-15 anti-Cy5 antibody is used to tag CyDye 5 for detection and quantitation and signal applification by or within immunocytochemical and immunohistochemical (IHC) techniques such as ELISA, immunoprecipitation, immunocytochemistry, dot blot assay, and protein microarrays. Anti-Cy5 antibody is an important tool for Cy5 signal amplification in assays where biomolecules are labeled with Cy5.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Talia D Valentini et al.
Nature communications, 11(1), 2287-2287 (2020-05-10)
Culture-independent studies of cystic fibrosis lung microbiota have provided few mechanistic insights into the polymicrobial basis of disease. Deciphering the specific contributions of individual taxa to CF pathogenesis requires comprehensive understanding of their ecophysiology at the site of infection. We
Eileen Sun et al.
Journal of virology, 87(5), 2673-2685 (2012-12-21)
As an obligate pathogen, influenza virus requires host cell factors and compartments to mediate productive infection and to produce infectious progeny virus. Recently, several small interfering RNA (siRNA) knockdown screens revealed influenza virus host dependency proteins, all of which identified
Dissecting the role of COPI complexes in influenza virus infection
Sun E, et al.
Journal of Virology, 87(5), 2673-2685 (2013)
Jiang He et al.
PLoS pathogens, 9(10), e1003701-e1003701 (2013-10-17)
As an obligatory pathogen, influenza virus co-opts host cell machinery to harbor infection and to produce progeny viruses. In order to characterize the virus-host cell interactions, several genome-wide siRNA screens and proteomic analyses have been performed recently to identify host
Masaaki Kuwajima et al.
PloS one, 15(1), e0226797-e0226797 (2020-01-16)
Analysis of neuronal compartments has revealed many state-dependent changes in geometry but establishing synapse-specific mechanisms at the nanoscale has proven elusive. We co-expressed channelrhodopsin2-GFP and mAPEX2 in a subset of hippocampal CA3 neurons and used trains of light to induce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service