S3762
Streptavidin−FITC from Streptomyces avidinii
essentially salt-free, lyophilized powder, ≥5 units/mg protein
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
conjugate
FITC conjugate
Quality Level
form
essentially salt-free, lyophilized powder
specific activity
≥5 units/mg protein
extent of labeling
3-9 mol FITC per mol streptavidin
storage temp.
−20°C
Application
The optimal working dilution should be determined empirically using a range of dilutions from a 1 mg/ml stock in buffer.
Unit Definition
One unit will bind 1 μg biotin.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Molecular therapy. Nucleic acids, 20, 231-241 (2020-03-21)
Esophageal squamous cell carcinoma (ESCC) is a common cancer occurring in males and females worldwide. Accumulating evidence continues to highlight the crucial roles of long non-coding RNAs (lncRNAs) in the process of tumorigenesis. However, the regulatory mechanism of lncRNAs in
Molecular therapy. Nucleic acids, 17, 644-656 (2019-08-11)
Colorectal cancer (CRC) is a frequently occurring lethal disorder with heterogeneous outcomes and drug responses. Recent studies have demonstrated that long non-coding RNAs (lncRNAs) play a critical role in carcinogenesis. Hence, the aim of this study was to investigate the
Tetrahydropteridines suppress gene expression and induce apoptosis of activated RAW264.7 cells via formation of hydrogen peroxide.
Free Radical Biology & Medicine, 37(3), 375-385 (2004)
The motion of a single molecule, the lambda-receptor, in the bacterial outer membrane.
Biophysical Journal, 83(6), 3152-3161 (2002)
Journal of cellular physiology, 235(2), 1141-1154 (2019-07-28)
Ovarian cancer (OC) is a fatal cancer in women, mainly due to its aggressive nature and poor survival rate. The lncRNA-miRNA-mRNA (long noncoding RNA-microRNA-messenger RNA) interaction is promising biomarkers for the improving prognosis of OC. Therefore, we explored the regulatory
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service