Skip to Content
Merck
All Photos(1)

Key Documents

P2714

Sigma-Aldrich

Protease Inhibitor Cocktail

lyophilized powder, for the inhibition of serine, cysteine and metallo-proteases, for general use, lyophilized powder

Synonym(s):

Protease Inhibitor Cocktail from bovine lung, Protease Inhibitor Mix, Protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
form:
lyophilized powder
solubility:
water: soluble
storage temp.:
−20°C

Product Name

Protease Inhibitor Cocktail powder, for general use, lyophilized powder

biological source

bovine lung

Quality Level

form

lyophilized powder

solubility

water: soluble

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The protease inhibitor cocktail has been optimized and tested for general use. It consists of water-soluble protease inhibitors that effectively inhibit serine, cysteine, and metalloproteases. This product is supplied as a lyophilized powder.

Specificity

Inhibits serine, cysteine, and metallo-proteases.

Application

Protease Inhibitor Cocktail powder has been used as a component:
  • in radioimmunoprecipitation assay buffer to lyse the avian crystalline lens samples for western blot analysis
  • in phosphate buffer solution (PBS) for grinding recombinant protein expressed leaves to purify virus-like particles (VLPs)
  • in PBS to dissolve lyophilized skin secretion sample of A. loloensis for peptide purification
  • in 0.1 M NaCl solution to wash and collect skin secretions of frog

Biochem/physiol Actions

The components of this mixture, such as AEBSF and Aprotinin act to inhibit serine proteases, including trypsin, chymotrypsin, and plasmin amongst others. Bestatin inhibits aminpeptidases. E-64 acts against cystein proteases. EDTA is an inhibitor of metalloproteases. Leupeptin acts against both serine and cystein proteases.

Components

This mixture contains individual components, including:

AEBSF at 2 mM
Aprotinin at 0.3 μM
Bestatin at 116 μ
E-64 at 14 μM
EDTA at 1 mM
Leupeptin at 1 μM

Each component has specific inhibitory properties.

Caution

The lyophilized powder is stable for 4 years when stored at -20°C.

Quantity

One bottle makes 100 mL of cocktail, which are sufficient to the inhibition of proteases present in approximately 20 gram of cell extract.

Not all extracts contain the same levels of endogenous enzymes, and it may be necessary to adjust the volume of cocktail required.

Preparation Note

This product is supplied as a lyophilized powder. One bottle prepares 100 mL of a cocktail solution.

Other Notes

For R&D use only. Not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safety and immunogenicity of plant-produced African horse sickness virus-like particles in horses
Dennis SJ, et al.
Veterninary Research, 49(1), 1-6 (2018)
Five novel antimicrobial peptides from skin secretions of the frog, Amolops loloensis
Wang M, et al.
Journal of biological and chemical chronicles, 155(1), 72-76 (2010)
F Maier et al.
Osteoarthritis and cartilage, 27(9), 1382-1391 (2019-05-24)
Given the structural changes associated with the progression of Osteoarthritis (OA), we hypothesized that patterns of through-thickness, large-strain shear evolve with early-stage OA. We therefore aimed to determine whether and how patterns of shear strains change during early-stage OA to
Céline Méhats et al.
PloS one, 3(10), e3445-e3445 (2008-10-23)
Prolonged neonatal exposure to hyperoxia is associated with high mortality, leukocyte influx in airspaces, and impaired alveolarization. Inhibitors of type 4 phosphodiesterases are potent anti-inflammatory drugs now proposed for lung disorders. The current study was undertaken to determine the effects
Giorgio Ramadori et al.
Nature communications, 10(1), 3545-3545 (2019-08-09)
Tens of millions suffer from insulin deficiency (ID); a defect leading to severe metabolic imbalance and death. The only means for management of ID is insulin therapy; yet, this approach is sub-optimal and causes life-threatening hypoglycemia. Hence, ID represents a

Articles

While aprotinin and bovine pancreatic trypsin inhibitor (BPTI) are the same protein sequence, the term aprotinin is typically used when describing the protein derived from bovine lung.

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service