Skip to Content
Merck
All Photos(1)

Documents

A4188

Sigma-Aldrich

N-Acetyl-Ile-Glu-Thr-Asp-7-Amido-4-methylcoumarin

≥97% (HPLC), powder

Synonym(s):

Ac-IETD-AMC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H41N5O12
Molecular Weight:
675.68
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white

solubility

DMSO/DMF: 1 mg/mL

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(C)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C31H41N5O12/c1-6-14(2)26(32-17(5)38)30(46)34-20(9-10-23(39)40)28(44)36-27(16(4)37)31(47)35-21(13-24(41)42)29(45)33-18-7-8-19-15(3)11-25(43)48-22(19)12-18/h7-8,11-12,14,16,20-21,26-27,37H,6,9-10,13H2,1-5H3,(H,32,38)(H,33,45)(H,34,46)(H,35,47)(H,36,44)(H,39,40)(H,41,42)/t14-,16+,20-,21-,26-,27-/m0/s1

InChI key

ZYTCBVQZQSFKHQ-HGZCCCDHSA-N

Amino Acid Sequence

NAc-Ile-Glu-Thr-Asp-AMC

Application

Fluorogenic substrate for caspase 8 and granzyme B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Fromigue et al.
British journal of cancer, 89(1), 178-184 (2003-07-03)
Bone tissue constitutes a fertile 'soil' for metastatic tumours, notably breast cancer. High concentrations of growth factors in bone matrix favour cancer cell proliferation and survival, and a vicious cycle settles between bone matrix, osteoclasts and cancer cells. Classically, bisphosphonates
Bernardo Nuche-Berenguer et al.
Biochimica et biophysica acta, 1862(6), 1122-1136 (2016-02-26)
In a recent study we explored Group-1-p21-activated kinases (GP.1-PAKs) in rat pancreatic acini. Only PAK2 was present; it was activated by gastrointestinal-hormones/neurotransmitters and growth factors in a PKC-, Src- and small-GTPase-mediated manner. PAK2 was required for enzyme-secretion and ERK/1-2-activation. In
Bernardo Nuche-Berenguer et al.
American journal of physiology. Gastrointestinal and liver physiology, 310(11), G1015-G1027 (2016-04-02)
In pancreatic acinar cells, the Src family of kinases (SFK) is involved in the activation of several signaling cascades that are implicated in mediating cellular processes (growth, cytoskeletal changes, apoptosis). However, the role of SFKs in various physiological responses such
Huili Zhu et al.
Scientific reports, 6, 26859-26859 (2016-06-01)
Glucagon-like peptide-1 (GLP-1) is an incretin hormone that increases glucose-dependent insulin secretion to reduce the glucose level. Liraglutide, a long-acting GLP-1 analogue, has been found to have neuroprotective action in various experimental models. However, the protective mechanisms of liraglutide in
Fabien J Cousin et al.
PloS one, 7(3), e31892-e31892 (2012-03-24)
Gastric cancer is one of the most common cancers in the world. The "economically developed countries" life style, including diet, constitutes a risk factor favoring this cancer. Diet modulation may lower digestive cancer incidence. Among promising food components, dairy propionibacteria

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service