Skip to Content
Merck
All Photos(2)

Documents

A9919

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

technique(s)

direct ELISA: 1:45,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
western blot: 1:100,000-1:200,000 using detecting actin in total cell extract of HeLa cells (5-10 μg per well)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Binds all rabbit Igs.
IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins.
Immunoglobulin G (IgG) belongs to the immunoglobulin family. It is widely expressed and consists of a gamma (γ) heavy chain in the constant (C) region. IgG is a monomer with a molecular weight of about 150 kDa. The structure of IgG constitutes two identical heavy chains and two identical light chains with molecular weight of 50 kDa and 25 kDa respectively. The primary structure of the antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1, γ2, γ3, and γ4, respectively. Limited digestion using papain cleaves the antibody into three fragments, two of which are identical and contain the antigen-binding activity. The third fragment does not possess antigen-binding activity and is known as fragment crystallizable (Fc). It interacts with cells and effector molecules. The Fc fragment contains the CH2 and CH3 domains of the antibody molecule.

Immunogen

purified rabbit IgG

Application

Citrullination of antithrombin by PADI4 was analyzed by ELISA using alkaline phosphatase conjugated goat anti-rabbit IgG as the secondary diluted at 1:5000 in 0.05M carbonate/bicarbonate buffer (Ph 9.6).
Goat Anti-Rabbit IgG (whole molecule)-Alkaline Phosphatase antibody has been used for immunohistochemistry and western blot applications. The antibody can also be used for direct ELISA at 1:45,000.
Lysates of human fibroblasts and Hela cells were analyzed by western blot using alkaline phosphatase-conjugated goat anti-rabbit IgG as the secondary antibody at a 1:30000 dilution.

Other Notes

Antibody adsorbed with human IgG.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byron H Hartman et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 236(10), 2875-2883 (2007-09-08)
Notch mediates the process of lateral inhibition that controls the production of hair cells in the inner ear. Hair cells are known to express Notch ligands Dll1 and Jag2, which signal through Notch1 in adjacent supporting cells. However, recent genetic
Ana do Vale et al.
Toxins, 9(11) (2017-11-15)
AIP56 (apoptosis-inducing protein of 56 kDa) is a key virulence factor of
Ling Lan et al.
Oncology letters, 14(6), 7733-7738 (2018-01-19)
All-trans-retinoic acid (ATRA) can enhance iodine uptake capability of thyroid tumors, but the mechanisms remain poorly understood. The aim of the present study was to investigate the effects of ATRA on isotope susceptibility, proliferation and invasion of anaplastic thyroid carcinoma
D P Gately et al.
Molecular biology of the cell, 9(9), 2361-2374 (1998-09-03)
Ataxia telangiectasia-mutated gene (ATM) is a 350-kDa protein whose function is defective in the autosomal recessive disorder ataxia telangiectasia (AT). Affinity-purified polyclonal antibodies were used to characterize ATM. Steady-state levels of ATM protein varied from undetectable in most AT cell
Christopher K Dugard et al.
Plant physiology, 171(3), 1905-1920 (2016-05-25)
Traditional marker-based mapping and next-generation sequencing was used to determine that the Arabidopsis (Arabidopsis thaliana) low cell wall arabinose mutant murus5 (mur5) encodes a defective allele of REVERSIBLY GLYCOSYLATED POLYPEPTIDE2 (RGP2). Marker analysis of 13 F2 confirmed mutant progeny from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service