Skip to Content
Merck
All Photos(1)

Key Documents

A-020

Supelco

Amobarbital solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O3
CAS Number:
Molecular Weight:
226.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(NC(C1(CC)CCC(C)C)=O)NC1=O

InChI

1S/C11H18N2O3/c1-4-11(6-5-7(2)3)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)

InChI key

VIROVYVQCGLCII-UHFFFAOYSA-N

General description

A Certified Spiking Solution® for amobarbital testing methods by GC/MS or LC/MS for clinical toxicology, medication monitoring, forensic analysis, pain prescription monitoring, or urine drug testing applications. Amobarbital is a barbiturate approved for treatment of anxiety, insomnia, and epilepsy.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Propofol, amobarbital… is it the substance that matters, or the question about the role of the Wada test in brain tumor patients?
Karl Schaller
World neurosurgery, 75(3-4), 428-430 (2011-05-24)
Daniel L Drane et al.
Epilepsia, 53(10), 1790-1798 (2012-07-12)
Exclusive right hemisphere language lateralization is rarely observed in the Wada angiography results of epilepsy surgery patients. Cortical stimulation mapping (CSM) is infrequently performed in such patients, as most undergo nondominant left hemisphere resections, which are presumed not to pose
Impact of pretreatment with amobarbital on electroconvulsive therapy outcomes.
Clint Ross et al.
The journal of ECT, 29(1), e10-e11 (2013-02-21)
M Zijlmans et al.
Journal of neurology, 259(8), 1632-1638 (2012-01-27)
The purpose of this study was to evaluate the contribution of posterior circulation to memory function by comparing memory scores between patients with and without a foetal-type posterior cerebral artery (FTP) during the intracarotid amobarbital procedure (IAP) in epilepsy patients.
Julián Carvajal-Castrillón et al.
Revista de neurologia, 55(9), 520-527 (2012-11-01)
An appropriate localization of ictal onset zone in refractory temporal lobe epilepsy favors an adequate outcome associated with surgical treatment. When video-electroencephalogram (video-EEG) and magnetic resonance imaging do not provide accurate data to locate ictal onset zone, the use of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service