Skip to Content
Merck
All Photos(1)

Documents

A1543

Sigma-Aldrich

Anti-Human IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

recombinant

expressed in goat

conjugate

alkaline phosphatase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:20,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot (chemiluminescent): 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains, with molecular weight of 50kDa and 25kDa respectively. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders. Goat anti-human IgG binds to human IgG. Immunoglobulin G (IgG) is part of the immunoglobulin family and is a widely expressed serum antibody. The primary structure of this antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains.

Immunogen

IgG purified from human serum

Application

Goat anti-human IgG can also be used for dot blot (1:30,000) and immunohistochemical (1:50) applications.
Serum human IgG levels were analyzed by solid phase ELISA assays using alkaline phosphatase-conjugated goat anti-human IgG antibody incubated for 1 hour at 37°C. Assays was developed using 0.1% p-nitrophenyl phosphate (Sigma) as the substrate. Antibodies to the Seoul (strain 80/39) virus was detected in human sera by western blot analysis using alkaline phosphatase-conjugated goat anti-human IgG antibody at a 1:1000 dilution. Blots were developed using nitroblue tetrazolium and 5-bromo-4-chloro-3-indoyl-phosphate substrate (Sigma).

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophana Chea et al.
Frontiers in immunology, 15, 1368066-1368066 (2024-05-16)
Aedes spp. are the most prolific mosquito vectors in the world. Found on every continent, they can effectively transmit various arboviruses, including the dengue virus which continues to cause outbreaks worldwide and is spreading into previously non-endemic areas. The lack
Heetae Lee et al.
Journal of medical virology, 84(12), 1943-1952 (2012-10-20)
A highly significant increase in anti-Vesivirus (family Caliciviridae) antibody prevalence, along the axis from healthy blood donors; donors with elevated transaminase; patients with clinical hepatitis; and patients with post-transfusion/dialysis hepatitis, has been reported in human sera from the USA and
Jessica E Manning et al.
The Journal of infectious diseases, 226(8), 1327-1337 (2021-11-01)
We established the first prospective cohort to understand how infection with dengue virus is influenced by vector-specific determinants such as humoral immunity to Aedes aegypti salivary proteins. Children aged 2-9 years were enrolled in the PAGODAS (Pediatric Assessment Group of
L Lledó et al.
Journal of medical microbiology, 51(10), 861-865 (2002-11-19)
Data relating to hantavirus infection in Spain are scarce and limited to rural areas. The aim of this work was to study the seroprevalence of hantavirus infection in the Autonomous Community of Madrid (ACM), a region containing both rural and
Jens Bukh et al.
Journal of medical virology, 80(1), 87-94 (2007-11-28)
GB virus-B (GBV-B) is the virus most closely related to hepatitis C virus (HCV). Thus, we have used GBV-B infection of tamarins, which develop acute hepatitis following experimental infection, as a surrogate model to study protective immunity. As challenge virus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service