Skip to Content
Merck
All Photos(4)

Key Documents

A3553

Sigma-Aldrich

Acrylamide

suitable for electrophoresis, ≥99% (HPLC), powder

Synonym(s):

2-Propenamide, Polyacrylamide, acrylic amide, akrylamid, Acrylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
Molecular Weight:
71.08
Beilstein:
605349
EC Number:
MDL number:
UNSPSC Code:
41105319
eCl@ss:
39031205
PubChem Substance ID:
NACRES:
NB.22

vapor density

2.45 (vs air)

Quality Level

vapor pressure

0.03 mmHg ( 40 °C)

Assay

≥99% (HPLC)

form

powder

technique(s)

electrophoresis: suitable

impurities

≤0.005% Acrylic acid
≤0.005% Insoluble in water

conductivity

≤10 μmho (35% solution)

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

solubility

water: soluble 2.5 g/10 mL, clear, colorless

cation traces

Fe: ≤1 ppm
Mg: ≤3 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acrylamide functions as a quencher of tryptophan fluorescence. Acrylamide along with bis-acrylamide crosslinks to form polyacrylamide gels, which is used to separate protein and other biomolecules. Bis-acrylamide is essential for crosslinking and acrylamide is required for the polymerization of monomers. Acrylamide alkylates SH groups of cysteine and ε-NH2 lysine residues of proteins during electrophoresis.

Application

Acrylamide has been used in the preparation of polyacrylamide gels.

Compatibility

This substance has been tested against several types of hand protection for CE compliance. See Related Products to find the recommended gloves for handling this product.

suggested gloves for splash protection

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Protein Protocols Handbook (2007)
Relationships among cell morphology, intrinsic cell stiffness and cell-substrate interactions
Chiang MYM, et al.
Biomaterials, 34(38), 9754-9762 (2013)
Chemistry, biochemistry, and safety of acrylamide. A review
Friedman M
Journal of Agricultural and Food Chemistry, 51(16), 4504-4526 (2003)
I-Chien Liao et al.
Advanced functional materials, 23(47), 5833-5839 (2014-03-01)
The development of synthetic biomaterials that possess mechanical properties that mimic those of native tissues remains an important challenge to the field of materials. In particular, articular cartilage is a complex nonlinear, viscoelastic, and anisotropic material that exhibits a very
Arianna Rath et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(39), 15668-15673 (2013-09-11)
SDS/PAGE is universally used in biochemistry, cell biology, and immunology to resolve minute protein amounts readily from tissue and cell extracts. Although molecular weights of water-soluble proteins are reliably determined from their SDS/PAGE mobility, most helical membrane proteins, which comprise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service