Skip to Content
Merck
All Photos(4)

Key Documents

A2765

Sigma-Aldrich

Azocasein

protease substrate, chromogenic, powder

Synonym(s):

Sulfanilamide-azocasein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

Product Name

Azocasein, protease substrate

biological source

bovine

form

powder

technique(s)

ligand binding assay: suitable

solubility

water: 5 mg/mL, clear, orange to very deep orange

ε (extinction coefficient)

≥25 at 440 nm in 0.1 M NaOH at 1%

storage temp.

2-8°C

General description

Azocasein is a chromogenic derivative of casein. Protease degrades azocasein to yield TCA-soluble azopeptides with high UV-absorbance. This azocasein assay is widely employed to estimate the protease production by bacterial fermentation on synthetic substrates from glucose and inorganic salts.

Application

Azocasein has been used as a substrate for determination of protease activity.
Azocasein is a nonspecific protease substrate. Hydrolysis of the casein releases the azo dye into the media where it is detected by absorbance at 440 nm.
Azocasein is an inflammatory agent that is used to induce amyloid A amylooidosis in experimental animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Oswaldo Gomez et al.
Molecular microbiology, 115(1), 84-98 (2020-09-09)
To overcome the metal restriction imposed by the host's nutritional immunity, pathogenic bacteria use high metal affinity molecules called metallophores. Metallophore-mediated metal uptake pathways necessitate complex cycles of synthesis, secretion, and recovery of the metallophore across the bacterial envelope. We
Hongfei Liu et al.
Protein and peptide letters, 27(11), 1102-1113 (2020-03-21)
Protein drugs have disadvantages, such as short half-lives, unstable biological activities, and low utilization efficiency. In this paper, a porous ion-responsive targeted drug delivery system was designed, combining biodegradable carriers with ion exchange technology to overcome problems for protein drug
Recombinant proteins from plants: production and isolation of clinically useful compounds, 3 (1998)
Isabel Morales-Belpaire et al.
Water research, 42(17), 4449-4456 (2008-09-03)
Laboratory data on the behaviour of the pathogenic form of the prion protein (PrP(Sc)) in environmental matrices such as sewage sludge is scarce. Direct experiments with this misfolded protein require strict safety measures, pathogen class-3 facilities and costly reagents. However
Yosuke Tashiro et al.
Journal of bacteriology, 191(24), 7509-7519 (2009-10-20)
The opportunistic human bacterial pathogen Pseudomonas aeruginosa produces membrane vesicles (MVs) in its surrounding environment. Several features of the P. aeruginosa MV production mechanism are still unknown. We previously observed that depletion of Opr86, which has a role in outer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service