Skip to Content
Merck
All Photos(1)

Key Documents

320617

Sigma-Aldrich

Ethyl 4-hydroxy-3-methoxycinnamate

98%

Synonym(s):

Ethyl ferulate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH=CHCO2C2H5
CAS Number:
Molecular Weight:
222.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

164-166 °C/0.5 mmHg (lit.)

mp

63-65 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)\C=C\c1ccc(O)c(OC)c1

InChI

1S/C12H14O4/c1-3-16-12(14)7-5-9-4-6-10(13)11(8-9)15-2/h4-8,13H,3H2,1-2H3/b7-5+

InChI key

ATJVZXXHKSYELS-FNORWQNLSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gururaj Joshi et al.
Neurochemistry international, 48(4), 318-327 (2006-01-03)
Ferulic acid ethyl ester (FAEE) is an ester derivative of ferulic acid, the latter known for its anti-inflammatory and antioxidant properties. Previous studies from our laboratory have shown that ferulic acid protects synaptosomal membrane system and neuronal cell culture systems
Yan Zheng et al.
Bioresource technology, 100(12), 2896-2901 (2009-03-04)
Feruloylated diacylglycerol (FDAG) was synthesized using a selective lipase-catalyzed the transesterification between ethyl ferulate and triolein. To optimize the reaction conversion and purity of FDAG, dual response surface was applied to determine the effects of five-level-five-factors and their reciprocal interactions
Liang Kong et al.
Analytical and bioanalytical chemistry, 386(2), 264-274 (2006-07-27)
A new screening and analysis method that combines in vitro metabolism with high-performance liquid chromatography-mass spectrometry (HPLC-MS) was developed for the screening and analysis of an antineoplastic compound, coniferyl ferulate, which is present in the rhizome of Rhizoma Chuanxiong. Infrared
Mirko Bunzel et al.
Journal of agricultural and food chemistry, 56(21), 10368-10375 (2008-10-10)
Valuable information about possible types of linkages, reaction mechanisms, and sequences for oxidative coupling of phenolic compounds in planta is available from in vitro model systems. Ferulate oligomers were generated in a system using ethyl ferulate, peroxidase, and hydrogen peroxide
Yun Feng et al.
Biochimica et biophysica acta, 1780(4), 659-672 (2008-01-31)
It is an important therapeutic strategy to protect mitochondria from oxidative stress, especially during ischemia-reperfusion. In the present study, an attempt has been made to evaluate the protective effects of caffeic acid phenethyl ester (CAPE) and its related phenolic compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service