Skip to Content
Merck
All Photos(2)

Documents

C0528

Sigma-Aldrich

Casein fluorescein isothiocyanate from bovine milk

Type III, essentially salt-free, lyophilized powder

Synonym(s):

FITC-casein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

type

Type III

form

essentially salt-free, lyophilized powder

extent of labeling

≥50 μg FITC per mg solid

solubility

water: 5 mg/mL, clear to hazy

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Casein fluorescein isothiocyanate from bovine milk has been used:
  • in accessing zona pellucida (ZP) and perivitelline space (PVS) to evaluate changes induced by protease treatment
  • to make a reference line in immunochromatographic devices
  • in running gel for the documentation of proteinases

highly sensitive protease substrate

Biochem/physiol Actions

Casein fluorescein isothiocyanate is mainly used as a substrate in estimating the protease activity of psychrotrophic bacteria. It is also used as a substrate for determining the activity of bacteria in milk.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunochromatographic assay for simple and rapid detection of Satsuma dwarf virus and related viruses using monoclonal antibodies
Kusano N, et al.
Journal of General Plant Pathology, 73(1), 66-71 (2007)
The supplementation of culture medium with protease improves the hatching rate of mouse embryos
Lee DR, et al.
Human Reproduction, 12(11), 2493-2498 (1997)
Characteristics of digestive enzymes of calanoid copepod species from different latitudes in relation to temperature, pH and food
Freese D, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 162(4), 66-72 (2012)
Emily M J Fennell et al.
Pharmacology research & perspectives, 10(4), e00993-e00993 (2022-08-06)
We recently described the identification of a new class of small-molecule activators of the mitochondrial protease ClpP. These compounds synthesized by Madera Therapeutics showed increased potency of cancer growth inhibition over the related compound ONC201. In this study, we describe
Tomasz Chamera et al.
Journal of molecular biology, 431(11), 2180-2196 (2019-04-27)
Hsp104 is a yeast chaperone that rescues misfolded proteins from aggregates associated with proteotoxic stress and aging. Hsp104 consists of N-terminal domain, regulatory M-domain and two ATPase domains, assembled into a spiral-shaped hexamer. Protein disaggregation involves polypeptide extraction from an

Articles

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Get better detection and quantification of proteases with this high-sensitivity red protease detection assay.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service