Skip to Content
Merck
All Photos(1)

Key Documents

A0664

Sigma-Aldrich

4-Aminobutylphosphonic acid

≥99%

Synonym(s):

P-(4-aminobutyl)-phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12NO3P
CAS Number:
Molecular Weight:
153.12
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Assay

≥99%

form

powder

color

white

application(s)

detection

SMILES string

NCCCCP(O)(O)=O

InChI

1S/C4H12NO3P/c5-3-1-2-4-9(6,7)8/h1-5H2,(H2,6,7,8)

InChI key

IDPXFPYGZRMMKZ-UHFFFAOYSA-N

Biochem/physiol Actions

4-Aminobutylphosphonic acid, a GABA B receptor ligand, is used in studies on the regulation of prolactin (PRL) secretion and differential GABA receptor research.
4-aminobutylphosphonic acid, the phosphonic acid analogue of delta-aminovaleric acid, inhibits gamma-aminobutyric acid B (GABA-B receptor) binding without influencing either isoproterenol- or forskolin-stimulated cyclic AMP production.

Linkage

Analog of δ-aminovaleric acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

L S Wong et al.
Pharmacology, biochemistry, and behavior, 38(4), 829-835 (1991-04-01)
The application of 1.2 and 12.0 micrograms/side of the GABAA receptor agonist 3-aminopropane sulphonic acid bilaterally into the nucleus accumbens (Acb) of rats nonsignificantly depressed locomotor activity as assessed in automated Animex activity cages, while the highest dose (60 micrograms/side)
Oktay Yildirim et al.
International journal of molecular sciences, 11(3), 1162-1179 (2010-05-19)
FePt nanoparticles (NPs) were assembled on aluminum oxide substrates, and their ferromagnetic properties were studied before and after thermal annealing. For the first time, phosph(on)ates were used as an adsorbate to form self-assembled monolayers (SAMs) on alumina to direct the
H P De Koning et al.
Endocrinology, 132(2), 674-681 (1993-02-01)
The activity of many endocrine cells is regulated by gamma-aminobutyric acid (GABA). The effects of GABA are mediated by GABAA and/or GABAB receptors. While GABAB receptors in the central nervous system have now been extensively characterized, little is known of
N Tian et al.
Brain research, 660(2), 267-274 (1994-10-17)
Amacrine and ganglion cells in the amphibian retina contain GABAB, as well as GABAA, receptors. Baclofen, a GABAB agonist, hyperpolarizes the dark membrane potential of these third order neurons and makes their light responses more transient. GABAB receptors in the
R M Woodward et al.
Molecular pharmacology, 43(4), 609-625 (1993-04-01)
Poly(A)+ RNA from mammalian retina expresses bicuculline/baclofen-insensitive gamma-aminobutyric acid (GABA) receptors in Xenopus oocytes with properties similar to those of homooligomeric GABA rho 1 receptors. The pharmacological profile of these rho-like receptors was extended by measuring sensitivities to various GABAA

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service