Skip to Content
Merck
All Photos(1)

Key Documents

69794

Sigma-Aldrich

Mineral oil

BioUltra, for molecular biology

Synonym(s):

White mineral oil (petroleum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12181504
NACRES:
NA.25
grade:
for molecular biology
technique(s):
PCR: suitable
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

product line

BioUltra

form

viscous liquid

technique(s)

PCR: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.467 (lit.)
n20/D 1.468

density

0.85 g/mL at 20 °C

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.7
λ: 280 nm Amax: 1.0

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Identification and characterization of two P450 enzymes from Citrus sinensis involved in TMTT and DMNT biosyntheses and Asian citrus psyllid defense.: This research identifies P450 enzymes involved in biosyntheses within Citrus sinensis, using mineral oil as a solvent for enzyme characterization, with implications for pest control strategies (Sun X et al., 2024).

  • Sustained Release of Salicylic Acid for Halting Peri-Implantitis Progression in Healthy and Hyperglycemic Systemic Conditions: A Gottingen Minipig Model.: The study investigates the sustained release of salicylic acid using mineral oil-based formulations to prevent peri-implantitis, showing potential for dental implant care in diverse health conditions (Bergamo ETP et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quan Hong Tran et al.
Genes, 11(3) (2020-03-12)
Induced point mutations are important genetic resources for their ability to create hypo- and hypermorphic alleles that are useful for understanding gene functions and breeding. However, such mutant populations have only been developed for a few temperate maize varieties, mainly
Janine K Reinert et al.
Frontiers in behavioral neuroscience, 13, 267-267 (2020-01-11)
Behavioral phenotyping of mice is often compromised by manual interventions of the experimenter and limited throughput. Here, we describe a fully automated behavior setup that allows for quantitative analysis of mouse olfaction with minimized experimenter involvement. Mice are group-housed and
Xuchu Ma et al.
Dalton transactions (Cambridge, England : 2003), (46)(46), 10250-10255 (2009-11-19)
Na(4)Si(4) and Na(4)Ge(4) are ideal chemical precursors for inorganic clathrate structures, clusters, and nanocrystals. The monoclinic Zintl phases, Na(4)Si(4) and Na(4)Ge(4), contain isolated homo-tetrahedranide [Si(4)](4-) and [Ge(4)](4-) clusters surrounded by alkali metal cations. In this study, a simple scalable route
Takeshi Itabashi et al.
Nature methods, 6(2), 167-172 (2009-01-20)
Accurate chromosome segregation during meiosis depends on the assembly of a microtubule-based spindle of proper shape and size. Current models for spindle-size control focus on reaction diffusion-based chemical regulation and balance in activities of motor proteins. Although several molecular perturbations
Benjamin Grau et al.
Purinergic signalling, 4(4), 357-364 (2008-09-09)
The formation of appropriate connections between neurons and their specific targets is an essential step during development and repair of the nervous system. Growth cones are located at the leading edges of the growing neurites and respond to environmental cues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service