Skip to Content
Merck
All Photos(1)

Key Documents

28609

Sigma-Aldrich

Cholesterol β-D-glucoside

≥97% (TLC)

Synonym(s):

Cholesteryl β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H56O6
CAS Number:
Molecular Weight:
548.79
MDL number:
UNSPSC Code:
12352211
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97% (TLC)

form

solid

optical activity

[α]/D -49.0±3.0°, c = 0.5 in pyridine

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O

InChI

1S/C33H56O6/c1-19(2)7-6-8-20(3)24-11-12-25-23-10-9-21-17-22(13-15-32(21,4)26(23)14-16-33(24,25)5)38-31-30(37)29(36)28(35)27(18-34)39-31/h9,19-20,22-31,34-37H,6-8,10-18H2,1-5H3/t20-,22+,23+,24-,25+,26+,27-,28-,29+,30-,31-,32+,33-/m1/s1

InChI key

FSMCJUNYLQOAIM-UQBZCTSOSA-N

Biochem/physiol Actions

A lipid mediator in heat stress responses in animals, shows anti-ulcer effect.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Potocka et al.
Acta biochimica Polonica, 55(1), 127-134 (2008-01-16)
A membrane-bound UDP-glucose : sterol glucosyltransferase from Solanum melongena (eggplant) leaves was partially purified and its specificity as well as molecular and kinetic properties were defined. Among a wide spectrum of 3-OH steroids (i.e. typical plant sterols, androstane, pregnane and
Gunthard Stübs et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(11), 3536-3544 (2010-02-10)
Lyme disease (LD) is the most common tick-borne disease in Europe, North America, and Asia. The etiologic agents of LD are spirochetes of the group Borrelia burgdorferi sensu lato, which possess a lipid content of 25-30% of the dry weight.
M Haque et al.
Journal of bacteriology, 178(7), 2065-2070 (1996-04-01)
The lipid and fatty acid profiles of eight Helicobacter spp. (H. nemestrinae, H. acinonyx, H. canis, Helicobacter sp. strain CLO-3, "H. rappini" [Flexispira rappini], H. pametensis, Helicobacter sp. strain Bird-B, and Helicobacter sp. strain Bird-C) and the fatty acid profiles
Salvador Peiru et al.
Biotechnology for biofuels, 8, 223-223 (2015-12-24)
Biodiesels produced from transesterification of vegetable oils have a major quality problem due to the presence of precipitates, which need to be removed to avoid clogging of filters and engine failures. These precipitates have been reported to be mostly composed
Ryan A Davis et al.
Chemical communications (Cambridge, England), 48(72), 9083-9085 (2012-08-03)
In a three-step protocol involving regioselective enzymatic acylation, per-O-trimethylsilylation, and a one-pot α-glycosidation-deprotection sequence, cholesteryl-6-O-tetradecanoyl-α-D-glucopyranoside (α-CAG) of Helicobacter pylori is afforded starting from glucose in an overall yield of 45%. The production of CAG can be scaled to make purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service