Skip to Content
Merck
All Photos(2)

Key Documents

89851

Supelco

β-Sitosterol β-D-glucoside

analytical standard

Synonym(s):

β-Daucosterol, Coriandrinol, Daucosterin, Eleutheroside A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H60O6
CAS Number:
Molecular Weight:
576.85
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥75.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)O[C@@H]5OC(CO)[C@@H](O)[C@H](O)C5O)C(C)C

InChI

1S/C35H60O6/c1-7-22(20(2)3)9-8-21(4)26-12-13-27-25-11-10-23-18-24(14-16-34(23,5)28(25)15-17-35(26,27)6)40-33-32(39)31(38)30(37)29(19-36)41-33/h10,20-22,24-33,36-39H,7-9,11-19H2,1-6H3/t21-,22-,24+,25+,26-,27+,28+,29-,30-,31+,32-,33-,34+,35-/m1/s1

InChI key

NPJICTMALKLTFW-OFUAXYCQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Sitosterol β-D-glucoside is a steroid glycoside having, β-sitosterol as the aglycone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: triticum withania serenoa

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

681.8 °F

Flash Point(C)

361 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-ming Sun et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(7), 1057-1059 (2011-11-10)
To study the chemical constituents of the aerial part of Ligusticum jeholense. The constituents were isolated by sillica gel column chromatography, Sephadex LH-20 column chromatography and their structures were elucidated by spectral analysis. Seven compounds were separated from the EtOH
Tang W and Eisenbrand G et al.
Chinese Drugs of Plant Origin: Chemistry, Pharmacology, and Use in Traditional and Modern Medicine (2013)
Qian-Liang Chen et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(8), 1214-1216 (2012-01-12)
To systematically study the chemical constituents in the roots of Gentiana dahurica. Various column chromatographic techniques were used for isolation and purification. The structures were elucidated on the basis of spectral data (UV, IR, MS, NMR) and identified by comparing
Huan-Kai Yao et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 34(5), 716-718 (2011-10-01)
To investigate the chemical constituents of Kalopanax septemlobus. Chromatographic techniques including silica gel, gel, semi-preparative HPLC and PTLC as well as recrystallization were employed in the isolation and purification, and the structures were elucidated by spectral analysis and physical and
Xin-Ping Cheng et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 33(11), 1727-1730 (2011-03-26)
To study the chemical constituents of the root tube from Pteroxygonum giraldii. Column chromatography and spectral analysis were used to isolate and identify the constituents. Ten compounds were isolated and identified as beta-sitosterol (I), beta-sitosterol glucoside (II), 4', 5,5', 7-tetrahydroxy-3'-methoxy-3'-O-alpha-L-arabinopyranosyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service