Skip to Content
Merck
All Photos(7)

Documents

793604

Sigma-Aldrich

Potassium bromide

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

anhydrous

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

pH

5-6 (25 °C, 119 g/L)

mp

734 °C (lit.)

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for removal of peripheral membrane proteins.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Puja Kumari et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 152, 109-118 (2015-07-22)
This paper reports the enhancement in photoluminescence due to the alkaline ions (M(2+)) co-doping in YVO4:Eu(3+) phosphors prepared by solid state reaction method. The as synthesized samples were found to have single phase tetragonal structure and the incorporation of dopant/codopant
Hitesh Kulhari et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1511-1520 (2015-05-03)
Docetaxel (DTX) is an anticancer drug that is used alone and in combination with other drugs to treat tumours. However, it suffers from the drawback of non-specific cytotoxicity. To improve the therapeutic potential of DTX, we report the synthesis of
Samy M El-Megharbel et al.
Chemico-biological interactions, 220, 169-180 (2014-07-08)
A new silver-carbocysteine (Ccy-Ag) complex [Ag2(Ccy)2(H2O)2] has been synthesized and characterized by using a combination of FTIR, Raman, molar conductivity, (1)H NMR, electronic spectra, thermal analyses, X-ray powder diffraction (XRD) and scanning electron microscopy (SEM). The infrared spectrum of Ccy-Ag
Zehua Yang et al.
Organic & biomolecular chemistry, 13(9), 2694-2702 (2015-01-15)
A new P-stereogenic PNP pincer-Pd complex was readily prepared from optically pure 2,6-bis[(boranato(tert-butyl)methylphosphino)methyl]pyridine. It was used in the asymmetric intramolecular hydroamination of amino-1,3-dienes, with the desired products being obtained in good yields and with excellent regioselectivities and up to moderate
Vijayan Thamilarasan et al.
European journal of medicinal chemistry, 89, 266-278 (2014-12-03)
Metal complexes of the type Mn(bpy)2(N3)2 (1), Co(bpy)2(N3)2·3H2O (2) and Zn2(bpy)2(N3)4 (3) (Where bpy = 2,2-bipyridine) have been synthesized and characterized by elemental analysis and spectral (FT-IR, UV-vis) studies. The structure of complexes (1-3) have been determined by single crystal X-ray diffraction

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service