Merck
  • P-stereogenic PNP pincer-Pd catalyzed intramolecular hydroamination of amino-1,3-dienes.
P-stereogenic PNP pincer-Pd catalyzed intramolecular hydroamination of amino-1,3-dienes.
Organic & biomolecular chemistry (2015-01-15)
Zehua Yang, Chao Xia, Delong Liu, Yangang Liu, Masashi Sugiya, Tsuneo Imamoto, Wanbin Zhang
ABSTRACT
A new P-stereogenic PNP pincer-Pd complex was readily prepared from optically pure 2,6-bis[(boranato(tert-butyl)methylphosphino)methyl]pyridine. It was used in the asymmetric intramolecular hydroamination of amino-1,3-dienes, with the desired products being obtained in good yields and with excellent regioselectivities and up to moderate enantioselectivities. The absolute configuration of one of the hydroamination products was determined by X-ray crystallography studies. This simple and efficient procedure can be used for the synthesis of allyl-type chiral pyrrolidine derivatives.
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis