Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium bromide
Synonym(s):
Potassium bromide
Linear Formula:
KBr
CAS Number:
7758-02-3
Molecular Weight:
119.00
EC Number:
231-830-3
Products
Product Category
salts (12)
solvents (1)
Brand
SAFC (1)
Sigma-Aldrich (9)
Supelco (3)
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (3)
ACS reagent (2)
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
221864
Potassium bromide,
≥99% trace metals basis
Expand
View Pricing
243418
Potassium bromide,
ACS reagent, ≥99.0%
Expand
View Pricing
1.04907
Potassium bromide,
for IR spectroscopy Uvasol
®
Expand
View Pricing
746444
Potassium bromide,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
1.04905
Potassium bromide,
for analysis (max. 0,000001% Hg) EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
P9881
Potassium bromide,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
60089
Potassium bromide,
BioUltra, ≥99.5% (AT)
Expand
View Pricing
449962
Potassium bromide,
anhydrous, powder, 99.999% trace metals basis
Expand
View Pricing
793604
Potassium bromide,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing
451010
Potassium bromide,
anhydrous, powder, 99.95% trace metals basis
Expand
View Pricing
1.04904
Potassium bromide,
99.999 Suprapur
®
Expand
View Pricing
1.04900
Potassium bromide,
EMPROVE
®
EVOLVE
Expand
View Pricing
P0838
Potassium bromide,
BioXtra, ≥99.0%
Expand
View Pricing