Skip to Content
Merck
All Photos(1)

Key Documents

72252

Sigma-Aldrich

Nickel(II) nitrate hexahydrate

puriss. p.a., ≥98.5% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
290.79
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.5% (KT)
form:
crystals or chunks

grade

puriss. p.a.

Quality Level

Assay

≥98.5% (KT)

form

crystals or chunks

mp

56 °C (lit.)

density

2.05 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤10 mg/kg
Co: ≤100 mg/kg
Cu: ≤20 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Na: ≤50 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

SMILES string

O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Ni.6H2O/c2*2-1(3)4;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;

InChI key

AOPCKOPZYFFEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) nitrate hexahydrate (Ni(NO3)2.6H2O) is a hydrated nickel(II) salt. Its thermal degradation has been investigated by thermogravimetric methods under quasi-isothermal conditions.

Application

Nickel(II) nitrate hexahydrate may be used as precursor for the synthesis of [Ni1/3Co1/3Mn1/3]O2 powders and Ni(II) oxide aerogels.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monolithic nickel (II)-based aerogels using an organic epoxide: the importance of the counterion.
Gash AE, et al.
Journal of Non-Crystalline Solids, 350, 145-151 (2004)
Marisa Freitas et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 26(1), 13-21 (2012-10-26)
Nickel is an ubiquitous transition metal that is industrially applied in many forms, which inevitably leads to a high degree of occupational and environmental exposure. Over-exposure to nickel can produce a variety of adverse effects on human health, including allergy
H Tanojo et al.
Acta dermato-venereologica. Supplementum, (212)(212), 19-23 (2002-01-23)
Allergic contact dermatitis due to nickel salts is common. It is therefore important to measure the permeation of these salts through the stratum corneum (SC), the primary rate-limiting domain in skin. An advanced diffusion system and analytical techniques now enable
J-C Amiard et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 147(3), 378-385 (2008-02-16)
In mussels impacted by the V- and Ni-rich oil dispersed in the environment after the < Erika > wreck, a significant and positive correlation was observed between metallothionein (MT) and V concentrations whereas no correlation was observed between MT and
Thermal decomposition of nickel nitrate hexahydrate, Ni(NO3)2?6H2O, in comparison to Co(NO3)2?6H2O and Ca(NO3)2?4H2O.
Brockner W, et al.
Thermochimica Acta, 456(1), 64-68 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service