Skip to Content
Merck
All Photos(7)

Key Documents

216828

Sigma-Aldrich

Iron(III) nitrate nonahydrate

greener alternative

ACS reagent, ≥98%

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
moist crystals

grade

ACS reagent

Assay

≥98%
98.0-101.0% (ACS specification)

form

moist crystals

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.005% insolubles
≤0.1% Substances not pptd. by NH4OH (as sulfates)

pH

1.5 (20 °C)

mp

47 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.01%
K: ≤0.005%
Mg: ≤0.005%
Na: ≤0.05%

greener alternative category

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) nitrate nonahydrate is an inorganic iron salt that is used as an iron precursor for the synthesis of iron oxide nanoparticles.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

A variety of organically templated iron phosphates, of interest because of their rich crystal chemistry and their ability to absorb large molecules in their micropores,were prepared using this compound.
Iron(III) nitrate nonahydrate can be used as:     
  • A catalyst in the oxidation of 1o, 2o, and benzylic alcohols to their corresponding carbonyl compounds in the presence of heteropoly acids.      
  • A promoter in the synthesis of polycrystalline boron nitride nanotubes (BNNTs) by a ball milling-annealing method.

It can also be used to synthesize:      
  • Iron-doped mesoporous silica used as an efficient catalyst for the degradation of rhodamine B dye under acidic conditions.
  • Iron(III)-doped titania nanoparticles by modified sol-gel method.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanostructured few-layer graphene with superior optical limiting properties fabricated by a catalytic steam etching process.
Sun Z, et al.
The Journal of Physical Chemistry C, 117(22), 11811-11817 (2013)
Ian S Metcalfe et al.
Nature chemistry, 11(7), 638-643 (2019-05-28)
All real processes, be they chemical, mechanical or electrical, are thermodynamically irreversible and therefore suffer from thermodynamic losses. Here, we report the design and operation of a chemical reactor capable of approaching thermodynamically reversible operation. The reactor was employed for
Silica supported ferric nitrate nonahydrate: selective oxidation of benzoins under mild conditions.
Paul AM, et al.
Synthetic Communications, 33(15), 2581-2584 (2003)
Thermal decomposition of Ni (II) and Fe (III) nitrates and their mixture.
Elmasry MAA, et al.
Journal of Thermal Analysis and Calorimetry, 52(2), 489-495 (1998)
Synthesis of iron (III)-doped titania nanoparticles and its application for photodegradation of sulforhodamine-B pollutant.
Kumbhar A and Chumanov G.
Journal of Nanoparticle Research, 7(4-5), 489-498 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service