Skip to Content
Merck
All Photos(4)

Key Documents

237973

Sigma-Aldrich

Aluminum nitrate nonahydrate

ACS reagent, ≥98%

Synonym(s):

Aluminium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
375.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

≥98%
98.0-102.0% (ACS specification)

form

crystals

reaction suitability

reagent type: catalyst
core: aluminum

impurities

≤0.005% insolubles

pH

2.5-3.5 (25 °C, 50 g/L)

mp

73 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ca: 0.005%
Fe: ≤0.002%
K: 0.002%
Mg: 0.001%
Na: 0.005%
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum nitrate nonahydrate is an aluminum salt that can be prepared by dissolving aluminum or aluminum hydroxide in dilute nitric acid. It is used in the preparation of insulating papers, transformer core laminates, and cathode-ray tube heating elements.

Application

Aluminum nitrate nonahydrate can be used as a catalyst to synthesize:
  • β-Amino carbonyl compounds via one-pot Mannich reaction.
  • Bis(indolyl)methanes via electrophilic substitution reaction of indoles with aromatic aldehydes.
  • 3,4-Dihydropyrimidinones via solvent-free Biginelli reaction.

Features and Benefits

  • Easily recoverable through simple phase separation
  • Reusable without a distinct loss of catalytic activity

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of thermal decomposition and ?-pyrolysis of aluminum nitrate nonahydrate [Al (NO3)3? 9H2O].
El-Shereafy E, et al.
J. Radioanal. Nucl. Chem., 237(1-2), 183-186 (1998)
Okada et al.
Journal of colloid and interface science, 219(1), 195-200 (1999-10-21)
Amorphous aluminosilicate xerogels with various chemical compositions were prepared by coprecipitation, and their surface acidity and hydrophilicity were investigated by NH(3) gas temperature programed desorption (TPD), water vapor adsorption-desorption isotherms, and (27)Al magic angle spinning nuclear magnetic resonance (MAS NMR).
Macro-micro relationship in nanostructured functional
Composites
Zanotto A, et al.
Express Polymer Letters, 6(5), 410-416 (2012)
Preparation of mullite fibers from aluminum isopropoxide-aluminum nitrate-tetraethylorthosilicate solutions by sol-gel method.
Song KC.
Materials Letters, 35(5), 290-296 (1998)
Effect of preparation conditions on the porous properties of coprecipitated Al2O3-SiO2 xerogels synthesized from aluminium nitrate nonahydrate and tetraethylorthosilicate.
Okada K, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 37(3), 355-364 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service