Skip to Content
Merck
All Photos(2)

Key Documents

538280

Sigma-Aldrich

Diethoxymethane

99.7%, contains 50-150 ppm BHT as stabilizer

Synonym(s):

Formaldehyde diethyl acetal, Diethoxymethane, Ethylal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(OC2H5)2
CAS Number:
Molecular Weight:
104.15
Beilstein:
1697253
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.7%
bp:
87-88 °C (lit.)
vapor pressure:
60 mmHg ( 25 °C)

vapor density

3.6 (vs air)

Quality Level

vapor pressure

60 mmHg ( 25 °C)

Assay

99.7%

form

liquid

autoignition temp.

346 °F

contains

50-150 ppm BHT as stabilizer

expl. lim.

8 %

refractive index

n20/D 1.373 (lit.)

bp

87-88 °C (lit.)

mp

-66.5 °C

density

0.831 g/mL at 25 °C (lit.)

SMILES string

CCOCOCC

InChI

1S/C5H12O2/c1-3-6-5-7-4-2/h3-5H2,1-2H3

InChI key

KLKFAASOGCDTDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethoxymethane (DEM) is a dialkoxymethane that can be synthesized by reacting ethanol and aqueous formaldehyde using macroporous cation-exchange resin Indion-130 as a catalyst. Infrared spectra of the conformers of DEM in various states have been obtained by matrix isolation infrared spectroscopy. The characteristic properties of DEM include low boiling point, low affinity for water, base stability and does not need drying for any reactions. Its infrared and Raman spectra have been reported.

Application

Diethoxymethane may be used as a substitute solvent to dichloromethane and toluene in the O-alkylation of different phenols in the presence of phase transfer catalysts (PTCs).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of ethanol and formaldehyde: use of versatile cation-exchange resins as catalyst in batch reactors and reactive distillation columns.
Chopade SP and Sharma MM.
Reactive functional Polymers, 32(1), 53-65 (1997)
Applications of diethoxymethane as a versatile process solvent and unique reagent in organic synthesis.
Boaz NW and Venepalli B.
Organic Process Research & Development, 5(2), 127-131 (2001)
Use of Diethoxymethane as a Solvent for Phase Transfer-Catalyzed O-Alkylation of Phenols.
Coleman MT and LeBlanc G.
Organic Process Research & Development, 14(3), 732-736 (2010)
Conformations of diethoxymethane: matrix isolation infrared and ab Initio studies.
Venkatesan V, et al.
The Journal of Physical Chemistry A, 107(39), 7727-7732 (2003)
Infrared and Raman Spectra of Diethoxymethane and Diethoxymethane-d2.
Nukada K.
Bulletin of the Chemical Society of Japan, 34(11), 1624-1630 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service