593230
Phenyllithium solution
1.9 M in dibutyl ether
Synonym(s):
PhLi
About This Item
Recommended Products
concentration
1.9 M in dibutyl ether
density
0.835 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
[Li]c1ccccc1
InChI
1S/C6H5.Li/c1-2-4-6-5-3-1;/h1-5H;
InChI key
NHKJPPKXDNZFBJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- 1-Acetyl-1′-diphenylphosphinoferrocene from ferrocenophane.
- Ethyl 2-acetylimino-1-benzyl-6-bromo-4-phenyl-1,4-dihydroquinoline-3-carboxylate from ethyl 1-benzyl-6-bromo-2-imino-1,2-dihydroquinoline-3-carboxylate.
- Ethyl 2-amino-1-benzoyl-6-bromo-4-phenyl-1,4-dihydroquinoline-3-carboxylate from (E)-ethyl 2-(acetylimino)-1-benzoyl-6-bromo-1,2-dihydroquinoline-3-carboxylate.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
77.0 °F - closed cup
Flash Point(C)
25 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service