Skip to Content
Merck
All Photos(2)

Key Documents

248568

Sigma-Aldrich

Calcium oxide

reagent grade

Synonym(s):

Lime, Quicklime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

form

powder

concentration

≥68.0% Ca (EDTA titration)

loss

10% loss on ignition, 1000°C (absorbed H2O and CO2)

pH

12.6 (20 °C)

bp

2850 °C (lit.)

density

3.3 g/mL at 25 °C (lit.)

SMILES string

O=[Ca]

InChI

1S/Ca.O

InChI key

ODINCKMPIJJUCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calcium oxide (CaO) is an alkaline earth metal oxide that has many industrial applications. Calcium oxide receives great attention in catalysis because it is affordable, environmentally friendly, and characterized by a strong basic nature, abundantly available, low methanol solubility and non-toxic compound.

Application

Calcium oxide is used:
  • As a catalyst during the transesterification of palm oil to produce biodiesel.
  • In the inactivation of Cryptosporidium spp. oocysts and Giardia spp. Cysts from WTS (water treatment sludge).
  • In combination with PAA (peracetic acid) to increases the biogas production potential of cow manure and maize straw.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ryo Kubota et al.
Biological engineering transactions, 4(2), 69-80 (2011-01-01)
We report on the use of a non-instrumented device for the implementation of a loop-mediated amplification (LAMP) based assay for the select-agent bacterial-wilt pathogen
Calcium oxide as a solid base catalyst for transesterification of soybean oil and its application to biodiesel production.
Kouzu M, et al.
Fuel: The Science and Technology of Fuel and Energy, 87(12), 2798-2806 (2008)
Preparation of biodiesel catalyzed by solid super base of calcium oxide and its refining process.
Huaping ZHU, et al.
Chinese Journal of Catalysis, 27(5), 391-396 (2006)
María Nazaret González-Alcaraz et al.
Journal of environmental management, 116, 72-80 (2013-01-08)
The aim of this study was to identify the effectiveness of liming in combination with vegetation for the recovery of slightly acidic, saline soils of eutrophic wetlands affected by mine wastes, under fluctuating flooding conditions. Simulated soil profiles were constructed
Sebastião Roberto Soares et al.
Waste management (New York, N.Y.), 33(1), 175-183 (2012-11-06)
The establishment of rules to manage Health Care Waste (HCW) is a challenge for the public sector. Regulatory agencies must ensure the safety of waste management alternatives for two very different profiles of generators: (1) hospitals, which concentrate the production

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service