Skip to Content
Merck
All Photos(1)

Key Documents

419494

Sigma-Aldrich

Osmium tetroxide

ACS reagent, ≥98.0%

Synonym(s):

Osmium(VIII)-oxide, ‘Osmic acid’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥98.0%
grade:
ACS reagent
form:
crystalline
solubility:
acetone: soluble(lit.)
carbon tetrachloride: soluble(lit.)
methyl tert-butyl ether: soluble(lit.)
toluene: soluble(lit.)
water: soluble(lit.)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

8.8 (vs air)

vapor pressure

7 mmHg ( 20 °C)

description

identification passes test

Assay

≥98.0%

form

crystalline

reaction suitability

reagent type: oxidant

clarity of soln

passes test

bp

130 °C (lit.)

mp

39.5-41 °C (lit.)

solubility

acetone: soluble(lit.)
carbon tetrachloride: soluble(lit.)
methyl tert-butyl ether: soluble(lit.)
toluene: soluble(lit.)
water: soluble(lit.)

storage temp.

2-8°C

SMILES string

O=[Os](=O)(=O)=O

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Osmium tetroxide may be used in the following processes:
  • As a tissue-staining contrast agent for staining tissues.[1]
  • Lactonization of alkenols to lactones by oxidation.[2]
  • Synthesis of the corresponding tetrols from 1,5-cycloocatdiene by bishydroxylation.[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the Stereoselectivity in Bisdihydroxylation of 1,5-cyclooctadiene with Osmium Tetroxide.
Kawazoe K, et al.
Synthetic Communications, 31(14), 2107-2112 (2001)
P Maupin-Szamier et al.
The Journal of cell biology, 77(3), 837-852 (1978-06-01)
We have studied the destruction of purified muscle actin filaments by osmium tetroxide (OsO4) to develop methods to preserve actin filaments during preparation for electron microscopy. Actin filaments are fragmented during exposure to OsO4. This causes the viscosity of solutions
Vacuum ultraviolet spectra of osmium tetroxide and ruthenium tetroxide.
Roebber JL, et al.
J. Chem. Phys. , 60(8), 3166-3173 (1974)
Direct Lactonization of Alkenols via Osmium Tetroxide-Mediated Oxidative Cleavage.
Schomaker JM, et al.
Organic Letters, 5(17), 3089-3092 (2003)
The crystal structure of osmium tetroxide.
Ueki T, et al.
Acta Crystallographica, 19(2), 157-160 (1965)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service