Skip to Content
Merck
All Photos(1)

Key Documents

75633

Sigma-Aldrich

Osmium tetroxide solution

suitable for electron microscopy, 2% in H2O

Synonym(s):

Osmium(IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
grade:
for analytical purposes
form:
solution
Pricing and availability is not currently available.

grade

for analytical purposes

form

solution

reaction suitability

reagent type: oxidant

concentration

2% in H2O

technique(s)

electron microscopy: suitable

color

colorless to yellow

density

1.0 g/mL at 20 °C
1.0 g/mL at 20 °C

application(s)

hematology
histology

storage temp.

2-8°C

SMILES string

O=[Os](=O)(=O)=O

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Osmium tetroxide (OsO4) also referred to as osmium, is a heavy tetrahedral, symmetrical molecule. It is both a dye and a non-coagulant fixative agent in electron microscopy primarily used for the examination of biological tissue components in the histopathology laboratory. Specimens are postfixed with osmium tetroxide to ensure the preservation of the lipids.

Application

Osmium tetroxide solution has been used as a fixative in electron microscopy.[1][2]

Biochem/physiol Actions

Osmium tetroxide is also referred to as osmium and is mainly used as a secondary fixative of tissues. The molecule reacts with lipid moieties and is responsible for the oxidation of unsaturated bonds of fatty acids. This reaction adds density and contrast to biological samples. In normal tissues, presence of osmium results in intense black staining which can be viewed by normal eye. In addition, it can be used as a mordant (a molecule which combines with stain or dye at later stages) in electron microscopy, thereby enhancing lead staining.[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-acetylpyridine-induced degeneration in the adult ascidian neural complex: Reactive and regenerative changes in glia and blood cells.
Medina BN, et al.
Developmental neurobiology, 75, 877-877 (2015)
Regulation of nitric-oxide production in hemocytes of the ascidian Phallusia nigra.
de Barros CM, et al.
Nitric Oxide, 38, 26-26 (2014)
Electron Microscopy: Principles and Techniques for Biologists (1999)
Bianca N S P Medina et al.
Developmental neurobiology, 75(8), 877-893 (2014-12-09)
Ascidians are interesting neurobiological models because of their evolutionary position as a sister-group of vertebrates and the high regenerative capacity of their central nervous system (CNS). We investigated the degeneration and regeneration of the cerebral ganglion complex of the ascidian
Carsten Købler et al.
Analytical and bioanalytical chemistry, 406(16), 3863-3873 (2014-01-23)
Ultrastructural characterisation is important for understanding carbon nanotube (CNT) toxicity and how the CNTs interact with cells and tissues. The standard method for this involves using transmission electron microscopy (TEM). However, in particular, the sample preparation, using a microtome to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service