Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Skip To

45359

Epoxy Embedding Medium kit

embedding resin for electron microscopy

Synonym(s):

Epon™ substitute embedding medium kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12171500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

pH

5-7

density

1.9 g/cm3

application(s)

hematology
histology

storage temp.

2-8°C

General description

The Epoxy Embedding Medium Kit is a very widely used embedding medium for electron microscopy. The embedding formulation originally published by Luft (1961) is excellent for both plant and animal tissues. Due to the low viscosity of the resin, it penetrates the tissue specimen faster than Araldite and other polymers. It hardens easily and uniformly at low temperatures with the addition of dodecenylsuccinic anhydride (DDSA), methyl nadic anhydride (MNA), and the accelerator 2,4,6-tris(dimethylaminomethyl)phenol (DMP-30). Slight shrinkage may occur during curing. This kit is useful for embedding a variety of tissues as a wide range of hardness can be obtained with this resin to suit a specific tissue type by using two different anhydride curing agents (DDSA and MNA).

Application

Epoxy Embedding Medium Kit has been used in the following applications:

  • to study the role of NDRG2, an essential colonic epithelial barrier regulator, in gut homeostasis maintenance and colitis-associated tumor development
  • to study the interaction of polymer scaffold nanofibers with growing cells
  • to study the morphological characterization of postembryonic development of blood–spleen barrier in ducks
  • to understand the role of NOD1/NOD2 receptors in Fusobacterium nucleatum-mediated neutrophil extracellular traps

Features and Benefits

  • Excellent medium for embedding a wide variety of tissues, including both plant and animal.
  • Facilitates rapid embedding (less than 3 hours).
  • Low viscosity facilitates rapid tissue penetration.
  • Desirable hardness is easy to obtain to suit a specific tissue type.

Other Notes

45345 Epoxy Embedding Medium 2x250 ml
45347 Hardener MNA 250 ml
45346 Hardener DDSA 250 ml
45348 Accelerator DMP 30 250 ml

Legal Information

Epon is a trademark of Hexion, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4534545348U4635
description

embedding resin for electron microscopy

description

suitable for microscopy

description

≥95% (NT)

description

white

density

1.9 g/cm3

density

-

density

0.969 g/mL at 25 °C (lit.)

density

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

pH

5-7

pH

-

pH

-

pH

-

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

room temp


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

275.0 °F

flash_point_c

135 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Should I first prepare Mixture A: and separately Mixture B: and then use these mixtures to infiltrate the tissue in ratios 1:1 and 1:2, or should I mix all four components together at once and then perform the infiltration? I am not clear about this step

    1 answer
    1. Mixture A is prepared by adding 5ml of the Epoxy Embedding Medium with 8 ml of DDSA. Mix well for at least 20 minutes. Mixture B is prepared by adding 8ml of Epoxy Embedding Medium with 7ml of NMA. Immediately before use mix 13 ml of Mixture A with 15 ml of Mixture B. Carefully add 16 drops of DPM-30 (1.5% to 2.0%). The DPM-30 (accelerator) should be added carefully since the block can become discolored if not measured properly and the block may be too brittle for satisfactory sectioning.
      Please refer to the usage protocol here:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/433/080/45359dat.pdf

      Helpful?

  2. Hallo, gelten nach dem Aushärten die gleichen H- und P-Sätze wie für das Kit?

    1 answer
    1. After curing, the hazard and precautionary statements will still apply to this kit.

      Helpful?

Reviews

No rating value

Active Filters