Skip to Content
Merck
All Photos(3)

Key Documents

402842

Sigma-Aldrich

Butyl acetate

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
technique(s):
tissue processing: suitable
bp:
124-126 °C (lit.)
vapor pressure:
15 mmHg ( 25 °C)
8 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

790 °F

expl. lim.

7.6 %

technique(s)

tissue processing: suitable

impurities

H2SO4, passes test (darkened)
≤0.0016 meq/g Titr. acid
≤0.1% water
≤0.2% C4H9OH

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5.3 g/L)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl acetate (BA, n-butyl acetate) is an ester commonly used as an organic solvent in cosmetic and pharmaceutical industries and as a flavoring agent in food industry.[1][2] One of the methods reported for its synthesis is the esterification reaction between acetic acid and butanol catalyzed by expandable graphite (EG).[1] The decomposition of butyl acetate by photocatalysis in gas phase has been studied using TiO2, Pt/TiO2 and WO3/TiO2 catalysts.[3] BA is a probable biofuel additive.[4]
Butyl acetate is a butyl ester of acetic acid. It exists in four isomeric forms: butyl acetate, sec-butyl acetate, isobutyl acetate and tert-butyl acetate. It is a colorless pleasant smelling liquid.[5]

Application

Butyl acetate has been used for the preparation of testicular tissue and cell cultures for immunohistochemical (IHC) and immunofluorescent (IF) staining.[6]
Butyl acetate may be used to abstract aluminium-hydroxyquinolate complex from soil extracts for determining aluminium spectrophotometrically and by fluorimetry.[7] It can also be used to abstract traces of gold from ores which is then determined by flame atomic absorption spectrometry.[8]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of trace gold in ores by butyl acetate extraction and flame atomic absorption spectrometry.
Shan XG and Lin YF.
Metallurgical Analysis, 3, 016-016 (2011)
Liquid-liquid equilibria for ternary systems acetic acid+ n-butyl acetate+ hydrocarbons at 293.15K.
Romain R, et al.
Fluid Phase Equilibria, 356, 264-270 (2013)
Catalytic Behaviors of Expansible Graphite in the Synthesis of Butyl Acetate.
Pang XY.
Journal of Chemistry, 9(4), 1816-1822 (2012)
Eagleson M.
Concise Encyclopedia Chemistry, 154-154 (1994)
The spectrophotometric and fluorometric determination of aluminum with 8-hydroxyquinoline and butyl acetate extraction.
Bloom PR, et al.
Soil Science Society of America Journal. Soil Science Society of America, 42(5), 713-716 (1978)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service