Skip to Content
Merck
All Photos(1)

Key Documents

W217514

Sigma-Aldrich

Isobutyl acetate

natural, ≥97%, FCC, FG

Synonym(s):

Acetic acid isobutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2175
Beilstein:
1741909
EC Number:
Council of Europe no.:
195c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.005
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

>4 (vs air)

vapor pressure

15 mmHg ( 20 °C)

Assay

≥97%

form

liquid

autoignition temp.

793 °F

expl. lim.

10.5 %

refractive index

n20/D 1.39 (lit.)

pH

6.7 (20 °C, 5 g/L)

bp

115-117 °C (lit.)

mp

−99 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; banana; fruity; ethereal; sweet

SMILES string

CC(C)COC(C)=O

InChI

1S/C6H12O2/c1-5(2)4-8-6(3)7/h5H,4H2,1-3H3

InChI key

GJRQTCIYDGXPES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Apple, banana, cocoa, fig, grape, grape brandy, pear, pineapple, sherry, strawberry and wine.

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - Pensky-Martens closed cup

Flash Point(C)

22 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 1441-1442 (1997)
Microbial production of scent and flavor compounds
Carroll AL, et al
Current Opinion in Biotechnology, 37, 8-15 (2016)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 1441-1442 (1997)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 103-103 (2003)
Bayram Yilmaz et al.
Reproductive toxicology (Elmsford, N.Y.), 22(4), 791-796 (2006-09-19)
Occupational exposure and sniffing of toluene-based organic solvents is an important public health problem. In this study, we have investigated the effects of paint thinner inhalation on testosterone synthesis and secretion in the male rat. A control group inhaled normal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service