Skip to Content
Merck
All Photos(3)

Key Documents

289159

Sigma-Aldrich

Cadmium acetate dihydrate

reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
266.53
Beilstein:
3730567
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.55

grade

reagent grade

Quality Level

Assay

98%

form

crystalline powder

reaction suitability

reagent type: catalyst
core: cadmium

SMILES string

[H]O[H].[H]O[H].CC(=O)O[Cd]OC(C)=O

InChI

1S/2C2H4O2.Cd.2H2O/c2*1-2(3)4;;;/h2*1H3,(H,3,4);;2*1H2/q;;+2;;/p-2

InChI key

AUIZLSZEDUYGDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - STOT RE 1 Oral

Target Organs

Kidney,Bone

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra de Souza Fonseca et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 5), m635-m636 (2012-05-17)
The reaction of cadmium acetate dihydrate with 2-acetyl-pyridine (4-phenyl-thio-semicarbazone) yielded the title compound, [Cd(C(14)H(13)N(4)S)(2)]. The Cd(II) atom is six-coordin-ated in a distorted octa-hedral environment by two deproton-ated thio-semicarbazone ligands acting in a tridentate chelating mode through two N and one
Jie Yang et al.
Journal of nanoscience and nanotechnology, 12(9), 7322-7328 (2012-10-06)
CdSe/Cd(1-x) Zn(x)S core/shell quantum dots (QDs) were fabricated in 1-octadecene via a two step synthesis. CdSe cores were first prepared using CdO, trioctylphosphine (TOP) selenium, and stearic acid. Subsquently, a Cd(1-x) Zn(x)S shell coating was carried out using zinc acetate
EPR single-crystal study of Cu2+ in cadmium acetate dihydrate: A seven-coordinated low-symmetry site.
Bonomo RP and Pilbrow JR.
Journal of Magnetic Resonance, 45(3), 404-417 (1981)
Mohammad Hossein Habibi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 133, 13-18 (2014-06-14)
ZnO@CdS core-shell nano-structure has been synthesized using zinc acetate dihydrate, and cadmium acetate dihydrate as simple precursors in a water-ethanol matrix without using any surfactant, ligand or chelating agents. The effect of different concentrations of cadmium acetate and sodium sulfide
Kanishka Biswas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(21), 6123-6129 (2007-05-15)
We have synthesized nanoparticles of hexagonal CdS in the diameter range 3-13 nm by the reaction of cadmium acetate dihydrate with thioacetamide in imidazolium [BMIM]-based ionic liquids. We have obtained three different particle sizes of CdS by changing the anion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service