Skip to Content
Merck
All Photos(3)

Key Documents

229490

Sigma-Aldrich

Cadmium acetate hydrate

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)2Cd · xH2O
CAS Number:
Molecular Weight:
230.50 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

powder, crystals or chunks

composition

Degree of hydration, 1-2

reaction suitability

core: cadmium
reagent type: catalyst

density

2.01 g/mL at 25 °C (lit.)

SMILES string

O.CC(=O)O[Cd]OC(C)=O

InChI

1S/2C2H4O2.Cd.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

KJNYWDYNPPTGLP-UHFFFAOYSA-L

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Melika Mahboub et al.
Nano letters, 16(11), 7169-7175 (2016-11-01)
Third generation photovoltaics are inexpensive modules that promise power conversion efficiencies exceeding the thermodynamic Shockley-Queisser limit, perhaps by using up- or down-converters, intermediate band solar cells, tandem cells, hot carrier devices, or multiexciton generation. Here, we report the efficient upconversion
Sanghwa Jeong et al.
Advanced healthcare materials, 7(24), e1800695-e1800695 (2018-11-20)
PbS/CdS core/shell quantum dots (QDs) that emit at the second near-infrared (NIR-II, 1000-1700 nm) window are synthesized. The PbS seed size and CdS shell thicknesses are carefully controlled to produce bright and narrow fluorescence that are suitable for multiplexing. A
Benjamin T Diroll et al.
Nature communications, 10(1), 4511-4511 (2019-10-06)
Colloidal quantum wells are two-dimensional materials grown with atomically-precise thickness that dictates their electronic structure. Although intersubband absorption in epitaxial quantum wells is well-known, analogous observations in non-epitaxial two-dimensional materials are sparse. Here we show that CdSe nanoplatelet quantum wells
Cai-Yu Lian et al.
Journal of inorganic biochemistry, 212, 111231-111231 (2020-08-31)
Autophagy dysregulation is implicated in cadmium (Cd)-induced nephrotoxicity. The mammalian target of rapamycin complex 1 (mTORC1) is a negative regulator of autophagy, but its role in Cd-induced autophagy inhibition and possible regulatory mechanisms remains poorly understood. In the present study
Phuong Le et al.
Nature communications, 10(1), 909-909 (2019-02-24)
The distribution of single-cell properties across a population of cells can be measured using diverse tools, but no technology directly quantifies the biochemical stimulation events regulating these properties. Here we report digital counting of growth factors in single cells using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service