Skip to Content
Merck
All Photos(3)

Key Documents

755087

Sigma-Aldrich

Cadmium(II) acetate

greener alternative

anhydrous, 99.995%

Synonym(s):

Cadmium(II) acetate, Acetic acid, cadmium salt, Bis(acetoxy)cadmium, Cadmium acetate, Cadmium diacetate, Cadmium ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(OCOCH3)2
CAS Number:
Molecular Weight:
230.50
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.995%

form

solid

reaction suitability

core: cadmium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

255-273 °C

greener alternative category

SMILES string

CC(=O)O[Cd]OC(C)=O

InChI

1S/2C2H4O2.Cd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

LHQLJMJLROMYRN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Cadmium(II) acetate can be used in the synthesis of cadmium oxide (CdO) thin films, which find usage in gas sensors, phototransistors, and diodes. It can also be used in the synthesis of cadmium sulfide (CdS) nanoparticles, which can be used in a variety of optoelectronic devices.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - STOT RE 1 Oral

Target Organs

Kidney,Bone

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of cadmium sulphide nanoparticles
Pattabi M and Uchil J
Solar Energy Materials and Solar Cells, 63(4), 309-314 (2000)
Spray deposition of highly transparent fluorine doped cadmium oxide thin films
Deokate RJ, et al.
Applied Surface Science, 254(7), 2187-2195 (2008)
R Saravanan et al.
Journal of colloid and interface science, 452, 126-133 (2015-05-04)
A ternary ZnO/Ag/CdO nanocomposite was synthesized using thermal decomposition method. The resulting nanocomposite was characterized by X-ray diffraction, field emission scanning electron microscopy, transmission electron microscopy, UV-Vis spectroscopy, and X-ray photoelectron spectroscopy. The ZnO/Ag/CdO nanocomposite exhibited enhanced photocatalytic activity under
Jennifer Thompson et al.
Reproductive toxicology (Elmsford, N.Y.), 20(4), 539-548 (2005-09-27)
The aim of this study was to examine cell death and cell proliferation in chick embryos destined to have ventral body wall defects as a result of cadmium (Cd) treatment. Embryos in shell-less culture were treated with 50 microL Cd
M Luisa Leret et al.
Toxicology, 186(1-2), 125-130 (2003-02-27)
The present study examines the effects of early simultaneous exposure to low level of lead and cadmium on anxiety-like behaviour in the rat, and on monoamine levels in the hypothalamus and hippocampus at weaning and adult animals. Rats were intoxicated

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service