T1253
2,4,6-Tris(2-pyridyl)-s-triazine
for spectrophotometric det. (of Fe), ≥98%
Synonym(s):
2,4,6-Tri(2-pyridyl)-s-triazine, TPTZ
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H12N6
CAS Number:
Molecular Weight:
312.33
Beilstein:
282581
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
grade
for spectrophotometric det. (of Fe)
Quality Level
Assay
≥98%
technique(s)
UV/Vis spectroscopy: suitable
mp
247-249 °C (lit.)
SMILES string
c1ccc(nc1)-c2nc(nc(n2)-c3ccccn3)-c4ccccn4
InChI
1S/C18H12N6/c1-4-10-19-13(7-1)16-22-17(14-8-2-5-11-20-14)24-18(23-16)15-9-3-6-12-21-15/h1-12H
InChI key
KMVWNDHKTPHDMT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2,4,6-Tris(2-pyridyl)-s-triazine is a complexing reagent used in spectrophotometric analysis.
Application
TPTZ may have been used in evaluating ionization constant and the formation constant of cobalt-TPTZ complex using spectrophotometric method.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Spectrophotometric determination of cobalt with 2, 4, 6-tris (2'-pyridyl)-s-triazine.
Janmohamed, Mirjehan J., and Gilbert H. Ayres.
Analytical Chemistry, 44.14, 2263-2268 (1972)
'True' ascorbic acid in plasma and serum: investigation of a new method.
W Samyn
Clinica chimica acta; international journal of clinical chemistry, 133(1), 111-117 (1983-09-15)
T Z Liu et al.
Clinical chemistry, 28(11), 2225-2228 (1982-11-01)
We describe a specific enzymatic spectrophotometric method for ascorbic acid in serum or plasma. Samples are analyzed indirectly by measuring the absorbance at 593 nm of a reaction product, a complex of ferrous ion and 2,4,6-tris(2-pyridyl)-s-triazine (Fe2+-TPTZ). This product is
D Ben-Shachar et al.
Journal of neurochemistry, 45(4), 999-1005 (1985-10-01)
Nutritional iron deficiency induced in rats causes a selective reduction of [3H]spiperone binding in caudate nucleus. This effect can be reversed by iron supplementation in vivo. The possibility that iron may be involved in the dopamine D2 receptor was investigated
Atomic-absorption spectrometric determination of copper(I) after adsorption of its 2,4,6-tri(2-pyridyl)-1,3,5-triazine complex with tetraphenylborate anion on microcrystalline naphthalene.
J L Lin et al.
The Analyst, 110(11), 1351-1354 (1985-11-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service